EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Effects of constituent ions of a phosphonium-based ionic liquid on molecular organization of H2O as probed by 1-propanol: tetrabutylphosphonium and trifluoroacetate ions.
PMID 26239281
Abstract
Aqueous solutions of tetrabutylphosphonium trifluoroacetate, [P4444]CF3COO, exhibit a liquid-liquid phase transition with a lower critical solution temperature. Herein, we characterized the constituent ions, [P4444](+) and CF3COO(-), in terms of their effects on the molecular organization of H2O on the basis of 1-propanol probing methodology devised by Koga et al. The resulting characterization of the hydrophobicity/hydrophilicity is displayed on a two-dimensional map together with previous results, for a total of four cations and nine anions of typical ionic liquid (IL) constituents. The results indicate that [P4444](+) is the most significant amphiphile with strong hydrophobic and equally strong hydrophilic contributions among the group of constituent cations of ILs studied so far. The hydration number for [P4444](+) was evaluated to be nH = 72, which is three times larger than that of a typical imidazolium-based cation, [C4mim](+). Self-aggregation of [P4444](+) was found to occur in an aqueous solution of [P4444]CF3COO above 0.0080 mole fraction of the IL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
132101
Sodium trifluoroacetate, 98%
C2F3NaO2
144800
Tetrabutylphosphonium chloride, 96%
C16H36ClP