EMAIL THIS PAGE TO A FRIEND
Oncology reports
miR-126 inhibits papillary thyroid carcinoma growth by targeting LRP6.
PMID 26239517
Abstract
microRNA-126 (miR-126) has been reported to play tumor suppressor roles in various types of cancers. Although it has been reported that miR-126 expression is downregulated in papillary thyroid carcinoma (PTC), the precise role and underlying molecular mechanism of miR-126 in PTC remains unclear. Therefore, the aims of the present study were to investigate the role and potential mechanism of miR-126 in tumorigenicity of PTC in vivo and in vitro. We observed that the miR-126 expression level was significantly downregulated in PTC tissue and PTC cell lines, the aberrant expression of miR-126 was correlated with lymph node metastasis, tumor size and TNM stage. We also showed that restoration of miR-126 in PTC cells inhibited cell proliferation, colony formations, migration and invasion, promoted cell apoptosis and cell cycle arrest at G1 stage in vitro, as well as inhibited tumor growth and decreased tumor volume and weight in vivo. Furthermore, low-density lipoprotein receptor‑related protein 6 (LRP6), a regulator of the Wnt/β‑catenin signaling cascade, was identified as a crucial target gene of miR-126. Overexpression of miR-126 inhibited LP6 expression on mRNA and protein levels, and deactivate Wnt/β-catenin signaling pathway. These results suggested that miR-126 functions as a tumor-suppressive miRNA by targeting LRP6 regulating Wnt/β-catenin signaling pathway and represents a therapeutic target for PTC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300742 Anti-LRP6 (C-term H1497) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300012
Anti-LRP6 (C-term T1546) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
L9419
Anti-LRP6 (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1300743 Anti-LRP6 (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300317 Anti-LRP6 (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
L9294
Anti-LRP6 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA029925
Anti-LRP6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501894
Anti-MMP-7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2500637
Anti-MMP7 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406133
Anti-MMP7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV46075
Anti-MMP7 antibody produced in rabbit, IgG fraction of antiserum
SAB1410789
Anti-MMP7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution