EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
The NF45/NF90 Heterodimer Contributes to the Biogenesis of 60S Ribosomal Subunits and Influences Nucleolar Morphology.
PMID 26240280
Abstract
The interleukin enhancer binding factors ILF2 (NF45) and ILF3 (NF90/NF110) have been implicated in various cellular pathways, such as transcription, microRNA (miRNA) processing, DNA repair, and translation, in mammalian cells. Using tandem affinity purification, we identified human NF45 and NF90 as components of precursors to 60S (pre-60S) ribosomal subunits. NF45 and NF90 are enriched in nucleoli and cosediment with pre-60S ribosomal particles in density gradient analysis. We show that association of the NF45/NF90 heterodimer with pre-60S ribosomal particles requires the double-stranded RNA binding domains of NF90, while depletion of NF45 and NF90 by RNA interference leads to a defect in 60S biogenesis. Nucleoli of cells depleted of NF45 and NF90 have altered morphology and display a characteristic spherical shape. These effects are not due to impaired rRNA transcription or processing of the precursors to 28S rRNA. Consistent with a role of the NF45/NF90 heterodimer in nucleolar steps of 60S subunit biogenesis, downregulation of NF45 and NF90 leads to a p53 response, accompanied by induction of the cyclin-dependent kinase inhibitor p21/CIP1, which can be counteracted by depletion of RPL11. Together, these data indicate that NF45 and NF90 are novel higher-eukaryote-specific factors required for the maturation of 60S ribosomal subunits.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA035775
Anti-D4S234E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2502092
Anti-FBL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502093
Anti-FBL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701663
Anti-fbl antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701789
Anti-fbl antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV40366
Anti-FBL antibody produced in rabbit, IgG fraction of antiserum
SAB4300633
Anti-Fibrillarin antibody produced in rabbit, affinity isolated antibody
SAB1406033
Anti-ILF2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007484
Anti-ILF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV35731
Anti-ILF2 antibody produced in rabbit, IgG fraction of antiserum
AV38968
Anti-ILF3 (AB1) antibody produced in rabbit, affinity isolated antibody
AV40523
Anti-ILF3 (AB2) antibody produced in rabbit, affinity isolated antibody
AV40522
Anti-ILF3 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB1406034
Anti-ILF3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001897
Anti-ILF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV38969
Anti-ILF3 antibody produced in rabbit, IgG fraction of antiserum
SAB2101154
Anti-ILF3 antibody produced in rabbit, affinity isolated antibody
SAB2106244
Anti-ILF3 antibody produced in rabbit, affinity isolated antibody
SAB2101297
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
HPA049830
Anti-KRAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107115
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB1305747
ANTI-KRAS2(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV46240
Anti-MRTO4 antibody produced in rabbit, IgG fraction of antiserum
HPA026446
Anti-MRTO4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB1410906
Anti-MRTO4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4502024
Anti-NPM antibody produced in rabbit, affinity isolated antibody
SAB4502025
Anti-NPM, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV34094
Anti-NPM1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV34095
Anti-NPM1 (AB2) antibody produced in rabbit, IgG fraction of antiserum