PloS one
A Triple Culture Model of the Blood-Brain Barrier Using Porcine Brain Endothelial cells, Astrocytes and Pericytes.
PMID 26241648
Abstract
In vitro blood-brain barrier (BBB) models based on primary brain endothelial cells (BECs) cultured as monoculture or in co-culture with primary astrocytes and pericytes are useful for studying many properties of the BBB. The BECs retain their expression of tight junction proteins and efflux transporters leading to high trans-endothelial electric resistance (TEER) and low passive paracellular permeability. The BECs, astrocytes and pericytes are often isolated from small rodents. Larger species as cows and pigs however, reveal a higher yield, are readily available and have a closer resemblance to humans, which make them favorable high-throughput sources for cellular isolation. The aim of the present study has been to determine if the preferable combination of purely porcine cells isolated from the 6 months old domestic pigs, i.e. porcine brain endothelial cells (PBECs) in co-culture with porcine astrocytes and pericytes, would compare with PBECs co-cultured with astrocytes and pericytes isolated from newborn rats with respect to TEER value and low passive permeability. The astrocytes and pericytes were grown both as contact and non-contact co-cultures as well as in triple culture to examine their effects on the PBECs for barrier formation as revealed by TEER, passive permeability, and expression patterns of tight junction proteins, efflux transporters and the transferrin receptor. This syngenic porcine in vitro BBB model is comparable to triple cultures using PBECs, rat astrocytes and rat pericytes with respect to TEER formation, low passive permeability, and expression of hallmark proteins signifying the brain endothelium (tight junction proteins claudin 5 and occludin, the efflux transporters P-glycoprotein (PgP) and breast cancer related protein (BCRP), and the transferrin receptor).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502981
Anti-Claudin 5, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4200538
Anti-Claudin-5 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200537
Anti-Claudin-5 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2100442
Anti-CLDN5 antibody produced in rabbit, affinity isolated antibody
SAB2501634
Anti-TJP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001636
Anti-TJP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102434
Anti-TJP1 antibody produced in rabbit, affinity isolated antibody
HPA001637
Anti-TJP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106392
Anti-TJP1 antibody produced in rabbit, affinity isolated antibody
SAB1306492
ANTI-ZO1(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C0378
Chloramphenicol, ≥98% (HPLC)
C11H12Cl2N2O5
C7795
Chloramphenicol, γ-irradiated
C11H12Cl2N2O5
C1919
Chloramphenicol, BioReagent, suitable for plant cell culture
C11H12Cl2N2O5
C0857
Chloramphenicol, meets USP testing specifications
C11H12Cl2N2O5
C1863
Chloramphenicol, Biotechnology Performance Certified, suitable for plant cell culture
C11H12Cl2N2O5
WH0007122M1
Monoclonal Anti-CLDN5 antibody produced in mouse, clone 3D8, purified immunoglobulin, buffered aqueous solution
S9133
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Na2O3Se
S5261
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Na2O3Se
214485
Sodium selenite, 99%
Na2O3Se