EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Crystal Growth, Structures, and Properties of the Complex Borides, LaOs2Al2B and La2Os2AlB2.
PMID 26241801
Abstract
Single crystals of two novel quaternary metal borides, LaOs2Al2B and La2Os2AlB2, have been grown from La/Ni eutectic fluxes. LaOs2Al2B crystallizes in tetragonal space group P4/mmm with the CeCr2Si2C-type structure, and lattice parameters a = 4.2075(6) Å and c = 5.634(1) Å. La2Os2AlB2 exhibits a new crystal structure in monoclinic space group C2/c with lattice parameters a = 16.629(3) Å, b = 6.048(1) Å, c = 10.393(2) Å, and β = 113.96(3)°. Both structures are three-dimensional frameworks with unusual coordination (for solid-state compounds) of the boron atoms by transition metal atoms. The boron atom is square planar in LaOs2Al2B, whereas it exhibits linear and T-shaped geometries in La2Os2AlB2. Electrical resistivity measurements reveal poor metal behavior (ρ300xa0K ∼ 900 μΩ cm) for La2Os2AlB2, consistent with the electronic band structure calculations, which also predict a metallic character for LaOs2Al2B.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266620
Boron, crystalline, 1 cm, 99.7% trace metals basis
B
333360
Boron, crystalline, −60 mesh, 99% trace metals basis
B
332445
Boron, amorphous, powder, (submicron)
B
15580
Boron, ≥95% (boron), amorphous powder
B
GF84487674
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF19976080
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF81914834
Boron, monofilament, 100m, diameter 0.1mm
B
GF54382888
Boron, monofilament, 10m, diameter 0.1mm
B
GF74687080
Boron, monofilament, 10m, diameter 0.2mm
B
GF40338132
Boron, monofilament, 5m, diameter 0.2mm
B
GF97861247
Boron, monofilament, 20m, diameter 0.1mm
B
GF04144586
Boron, monofilament, 20m, diameter 0.2mm
B
GF32655315
Boron, monofilament, 50m, diameter 0.1mm
B
GF70375833
Boron, monofilament, 50m, diameter 0.2mm
B
GF98835069
Boron, monofilament, 5m, diameter 0.1mm
B
GF39354048
Boron, monofilament, 200m, diameter 0.1mm
B
261130
Lanthanum, ingot (under oil), 99.9% trace rare earth metals basis
La
263117
Lanthanum, pieces
La
263109
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
La
GF13609111
Lanthanum, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
La
GF54212914
Lanthanum, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
La
GF43011391
Lanthanum, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
La
GF15415691
Lanthanum, foil, 100x100mm, thickness 0.1mm, as rolled, 99%
La
GF09165786
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
La
GF88400189
Lanthanum, foil, 29x33mm, thickness 0.5mm, as rolled, 99%
La
GF60507019
Lanthanum, rod, 50mm, diameter 6.35mm, cast, 99%
La
GF76465176
Lanthanum, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
La
GF45723326
Lanthanum, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
La
GF90307144
Lanthanum, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
La
GF35079554
Lanthanum, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
La