EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
The histone demethylase KDM1A is essential for the maintenance and differentiation of spermatogonial stem cells and progenitors.
PMID 26243864
Abstract
Little is known of the fundamental processes governed by epigenetic mechanisms in the supplier cells of spermatogenesis, the spermatogonial stem cells (SSCs). The histone H3 lysine demethylase KDM1A is expressed in spermatogonia. We hypothesized that KDM1A serves in transcriptional regulation of SSCs and fertility. Using a conditional deletion of Kdm1a [conditional knockout (cKO)] in mouse spermatogonia, we determined that Kdm1a is essential for spermatogenesis as adult cKO males completely lack germ cells. Analysis of postnatal testis development revealed that undifferentiated and differentiating spermatogonial populations form in Kdm1a-cKO animals, yet the majority fail to enter meiosis. Loss of germ cells in the cKO was rapid with none remaining by postnatal day (PND) 21. To gain insight into the mechanistic implications of Kdm1a ablation, we isolated PND 6 spermatogonia enriched for SSCs and analyzed their transcriptome by RNA sequencing. Loss of Kdm1a was associated with altered transcription of 1206 genes. Importantly, differentially expressed genes between control and Kdm1a-cKO animals included those that are essential for SSC and progenitor maintenance and spermatogonial differentiation. The complete loss of fertility and failure to establish spermatogenesis indicate that Kdm1a is a master controller of gene transcription in spermatogonia and is required for SSC and progenitor maintenance and differentiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407168
Anti-AOF2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1300533
Anti-DDX4 (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300532
Anti-DDX4 (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1407552
Anti-DDX4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037763
Anti-DDX4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA037764
Anti-DDX4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306220 ANTI-LSD1 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3500229
Anti-LSD1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500953
Anti-LSD1 antibody produced in rabbit, affinity isolated antibody
SAB1306581
ANTI-LSD1(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306580
ANTI-LSD1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306016
ANTI-PLZF(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406879
Anti-RAPGEF5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4503700
Anti-RAPGEF5 antibody produced in rabbit, affinity isolated antibody
SAB2700762
Anti-SYCP1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA021083
Anti-SYCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102345
Anti-SYCP1 antibody produced in rabbit, affinity isolated antibody
SAB2102346
Anti-SYCP3 antibody produced in rabbit, affinity isolated antibody
SAB1401670
Anti-SYCP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA043938
Anti-SYCP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039635
Anti-SYCP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001499
Anti-ZBTB16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300115
Monoclonal Anti-DDX4 antibody produced in mouse, clone 2F9H5, ascites fluid
L7293
Monoclonal Anti-LSD1 (AOF2) antibody produced in mouse, ~2 mg/mL, clone LSD1-12, purified immunoglobulin, buffered aqueous solution
SAB1402485
Monoclonal Anti-RAPGEF5 antibody produced in mouse, clone 2E4, purified immunoglobulin, buffered aqueous solution
WH0007704M1
Monoclonal Anti-ZBTB16 antibody produced in mouse, clone 3A7, purified immunoglobulin, buffered aqueous solution
SAB1404550
Monoclonal Anti-ZBTB16 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
SAB5300356
Monoclonal Anti-ZBTB16 antibody produced in mouse, clone 5B3, ascites fluid