PloS one
Intraductal Radiofrequency Ablation Followed by Locoregional Tumor Treatments for Treating Occluded Biliary Stents in Non-Resectable Malignant Biliary Obstruction: A Single-Institution Experience.
PMID 26244367
Abstract
To determine the safety and feasibility of intraductal radiofrequency ablation (RFA) followed by locoregional tumor treatments in patients with non-resectable malignant biliary obstruction and stent re-occlusion. Fourteen patients with malignant biliary obstruction and blocked metal stents were studied retrospectively. All had intraductal RFA followed by locoregional tumor treatments and were monitored clinically and radiologically. The practicality, safety, postoperative complications, jaundice remission, stent patency and survival time were analyzed. Combination treatment was successful for all patients. There were no severe complications during RFA or local treatments. All patients had stent patency restored, with a decline in serum bilirubin. Three patients had recurrent jaundice by 195, 237 and 357 days; two patients underwent repeat intraductal RFA; and one required an internal-external biliary drain. The average stent patency time was 234 days (range 187-544 days). With a median follow-up of 384 days (range 187-544 days), six patients were alive, while eight had died. There was no mortality at 30 days. The 3, 6, 12 and 18 month survival rates were 100%, 100%, 64.3% and 42.9%, respectively. Intraductal RFA followed by locoregional tumor treatments for occluded metal stents is safe and practically feasible and potential increase stent patency and survival times.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8624
Pirarubicin, ≥95% (HPLC)
C32H37NO12