EMAIL THIS PAGE TO A FRIEND
eLife
An extrafollicular pathway for the generation of effector CD8(+) T cells driven by the proinflammatory cytokine, IL-12.
PMID 26244629
Abstract
The proinflammatory cytokine IL-12 drives the generation of terminally differentiated KLRG1(+) effector CD8(+) T cells. Using a Toxoplasma vaccination model, we delineate the sequence of events that naïve CD8(+) T cells undergo to become terminal effectors and the differentiation steps controlled by IL-12. We demonstrate that direct IL-12 signaling on CD8(+) T cells is essential for the induction of KLRG1 and IFN-γ, but the subsequent downregulation of CXCR3 is controlled by IL-12 indirectly through the actions of IFN-γ and IFN-γ-inducible chemokines. Differentiation of nascent effectors occurs in an extrafollicular splenic compartment and is driven by late IL-12 production by DCs distinct from the classical CD8α(+) DC. Unexpectedly, we also found extensive proliferation of both KLRG1(-) and KLRG1(+) CD8(+) T cells in the marginal zone and red pulp, which ceases prior to the final KLRG1(Hi) CXCR3(Lo) stage. Our findings highlight the notion of an extrafollicular pathway for effector T cell generation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400378
Anti-KLRG1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401514
Anti-KLRG1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2700888
Anti-KLRG1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA051403
Anti-KLRG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501783 Anti-PREP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104775
Anti-PREP antibody produced in rabbit, affinity isolated antibody
HPA031388
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031389
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031390
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104515
Anti-PREP, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1401302
Monoclonal Anti-PREP antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution