EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
A high performance liquid chromatography with ultraviolet method for Eschweilera nana leaves and their anti-inflammatory and antioxidant activities.
PMID 26246741
Abstract
Eschweilera nana Miers is a tree widely distributed in Cerrado, Brazil. In this study, we aimed to describe its phytochemical properties and antioxidant and topical anti-inflammatory effects for the first time, as well validate an high performance liquid chromatography with ultraviolet/visible (HPLC-UV-Vis) method for the separation and quantification of the main components (hyperoside and rutin) in the hydroalcoholic extract of E. nana leaves. Structural identification of compounds in E. nana extract was performed by analysis of spectral data by (1)H nuclear magnetic resonance, (13)C nuclear magnetic resonance and/or ESI/EM. The HPLC-UV-Vis method was validated according International Conference on Harmonization (ICH) parameters. The 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS) and 1,1-diphenyl-2-picrylhydrazyl (DPPH) method were used for determination of in vitro antioxidant activities and the croton oil-induced inflammation for evaluation of in vivo anti-inflammatory effects. Hyperoside, rutin, α-amirin, β-amirin, β-sitosterol, and stigmasterol were identified in the hydroalcoholic extract of E. nana leaves. HPLC-UV-Vis was validated according to ICH parameters. Furthermore, in vitro and in vivo assays demonstrated that the hydroalcoholic extract and methanol fraction showed significant antioxidant and topical anti-inflammatory effects, as they were able to reduce ear edema induced by croton-oil application. This research showed the first phytochemical study of E. nana extract and their biological activities may be associated with the presence of flavonoids in the extracts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83388
Quercetin 3-D-galactoside, ≥97.0% (HPLC)
C21H20O12