EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Ensiling of seaweed for a seaweed biofuel industry.
PMID 26253914
Abstract
Effective biogas production from seaweed necessitates harvest at times of peak quality of biomass and low-loss preservation for year-around supply. Ensiling of five seaweed species and storage up to 90days was investigated as a method to preserve the methane yield potential. Adequate acidification by natural lactic acid fermentation was difficult due to low rapidly fermentable carbohydrate contents, high buffering capacities and low initial numbers of lactic acid bacteria. Nevertheless, products of silage fermentation increased methane yields by up to 28% and compensated for volatile solid losses during ensiling. Preservation of the original methane yield potential was achieved for four of five seaweed species, provided that silage effluent is collected and utilised. 10-28% of the ensiled biomass was released as effluent with methane yields of 218-423LNkg(-1) VS. If further optimised, ensiling represents an effective method of preservation crucial for an efficient seaweed biofuel industry.