EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
A novel humanized anti-HER2 antibody conjugated with MMAE exerts potent anti-tumor activity.
PMID 26253944
Abstract
Human epidermal growth factor receptor-2 (HER2) is a validated therapeutic target for breast cancer and trastuzumab (Herceptin), a humanized anti-HER2 antibody, has significant anti-cancer effects in the clinic. However, breast cancer patients often experience disease progression after prolonged Herceptin treatment. To develop a more effective therapy, we generated humanized monoclonal antibody hertuzumab and hertuzumab-drug conjugates as novel breast cancer therapies. The hertuzumab was conjugated with small molecule cytotoxic agents monomethylauristatin E (MMAE) or monomethylauristatin F (MMAF) with various linkers to generate antibody-drug conjugates (ADCs), which were evaluated for their in vitro and in vivo anti-cancer activities. Among these ADCs, hertuzumab-vc-MMAE can be effectively internalized and potently kill HER2 over-expressing tumor cells. In xenograft tumor models, hertuzumab-vc-MMAE showed a more potent anti-tumor activity than T-DM1, a FDA-approved ADC drug. More importantly, this novel ADC drug also showed superior anti-tumor activity than T-DM1 in trastuzumab- and lapatinib-resistant xenograft tumor models, suggesting its potential as an improved therapy for HER2-positive breast cancers. The novel ADC, hertuzumab-vc-MMAE, is an effective and selective agent for the treatment of HER2-positive breast tumors.