EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Sirtuin 1 (SIRT1) Deacetylase Activity and NAD⁺/NADH Ratio Are Imperative for Capsaicin-Mediated Programmed Cell Death.
PMID 26255724
Abstract
Capsaicin is considered a chemopreventive agent by virtue of its selective antigrowth activity, commonly associated with apoptosis, against cancer cells. However, noncancerous cells possess relatively higher tolerance to capsaicin, although the underlying mechanism for this difference remains unclear. Hence, this study aimed to elucidate the differential effects of capsaicin on cell lines from lung tissues by addressing the signal pathway leading to two types of cell death. In MRC-5 human fetal lung cells, capsaicin augmented silent mating type information regulation 1 (SIRT1) deacetylase activity and the intracellular NAD(+)/NADH ratio, decreasing acetylation of p53 and inducing autophagy. In contrast, capsaicin decreased the intracellular NAD(+)/NADH ratio, possibly through inhibition of tumor-associated NADH oxidase (tNOX), and diminished SIRT1 expression leading to enhanced p53 acetylation and apoptosis. Moreover, SIRT1 depletion by RNA interference attenuated capsaicin-induced apoptosis in A549 cancer cells and autophagy in MRC-5 cells, suggesting a vital role for SIRT1 in capsaicin-mediated cell death. Collectively, these data not only explain the differential cytotoxicity of capsaicin but shed light on the distinct cellular responses to capsaicin in cancerous and noncancerous cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1305642
ANTI-APG5 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
A0731
Anti-ATG5 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody
A0856
Anti-ATG5 (N-terminal) antibody produced in rabbit, affinity isolated antibody, PBS solution
GW22210F
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500448
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA042973
Anti-ATG5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3347
Anti-Bak antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B5897
Anti-Bak antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2500149
Anti-BAK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2100202
Anti-BAK1 antibody produced in rabbit, affinity isolated antibody
SAB1405470
Anti-BIRC5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
S5313
Anti-Sir2 (AS-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S5322
Anti-Sirt1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
S8191
Anti-Survivin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500269
Anti-Survivin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501460
Anti-Survivin antibody produced in rabbit, affinity isolated antibody
WH0009474M1
Monoclonal Anti-ATG5 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
A2859
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
SAB1404726
Monoclonal Anti-ATG5, (C-terminal) antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
SAB1402700 Monoclonal Anti-BAK1, (C-terminal) antibody produced in mouse, clone 1E4, ascites fluid
SAB4100184
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-B129, culture supernatant
SAB4100208
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-A129, culture supernatant
SAB5300513 Monoclonal Anti-BECN1 antibody produced in mouse, clone 2A4, ascites fluid
WH0000332M1
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 5B10, purified immunoglobulin, buffered aqueous solution
SAB4100131
Monoclonal Anti-BIRC5 antibody produced in mouse, clone BIRC5-A113, culture supernatant
SAB5300510 Monoclonal Anti-BIRC5 antibody produced in mouse, clone 2H5H2, ascites fluid
SAB5300511
Monoclonal Anti-BIRC5 antibody produced in mouse, clone 1H5, purified immunoglobulin, buffered aqueous solution
SAB1404972
Monoclonal Anti-SIRT1, (C-terminal) antibody produced in mouse, clone 7C2, purified immunoglobulin, buffered aqueous solution