EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Photo-responsive shell cross-linked micelles based on carboxymethyl chitosan and their application in controlled release of pesticide.
PMID 26256378
Abstract
An amphiphilic carboxymethyl chitosan conjugate with photolabile 2-nitrobenzyl side groups (NBS-CMCS) was synthesized, which could self-assemble into polymeric micelles in aqueous condition following by adding dropwise dialdehyde to form a cross-linking structure. TEM and (1)H NMR confirmed that the cross-linked micelles had a core-shell configuration with an average diameter of 140 ± 5.5 nm. DLS and TEM observations showed that the cross-linked micelles were stable in aqueous solution at pH 7.0 without light irradiation, while they could transfer into nanocapsules upon exposure to 365 nm UV light. Diuron, a photosynthetic inhibitor, was encapsulated in the cross-linked micelles reaching encapsulation efficiency as much as 91.9%. No release of the encapsulated diuron was detected without light, whereas a release rate as high as 96.8% over 8h at pH 7.0 buffer was observed under solar stimulated irradiation, indicating that the cross-linked micelles may be used as a photo-controlled sustained release carrier for the delivery of photosynthetic inhibitor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
107700
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
C7H10N2
714844
4-(Dimethylamino)pyridine solution, 0.5 M in THF
C7H10N2
714720
4-(Dimethylamino)pyridine solution, 0.5 M in ethyl acetate
C7H10N2
700169
4-(Dimethylamino)pyridine, ChemDose tablets, Loading: 0.04 mmol tablet.
C7H10N2
G5882
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
C5H8O2
G7651
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7776
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7526
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G6257
Glutaraldehyde solution, Grade II, 25% in H2O
C5H8O2
G6403
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
C5H8O2
340855
Glutaraldehyde solution, 50 wt. % in H2O
C5H8O2
49626
Glutaraldehyde solution, for electron microscopy, ~25% in H2O
C5H8O2
G4004
Glutaraldehyde solution, 25 wt. % in H2O
C5H8O2
W512303
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
C5H8O2
130672
N-Hydroxysuccinimide, 98%
C4H5NO3
239690
Succinic anhydride, ≥99% (GC)
C4H4O3