EMAIL THIS PAGE TO A FRIEND
Nature communications
Oestrogen sulfotransferase ablation sensitizes mice to sepsis.
PMID 26259151
Abstract
Sepsis is the host's deleterious systemic inflammatory response to microbial infections. Here we report an essential role for the oestrogen sulfotransferase (EST or SULT1E1), a conjugating enzyme that sulfonates and deactivates estrogens, in sepsis response. Both the caecal ligation and puncture (CLP) and lipopolysaccharide models of sepsis induce the expression of EST and compromise the activity of oestrogen, an anti-inflammatory hormone. Surprisingly, EST ablation sensitizes mice to sepsis-induced death. Mechanistically, EST ablation attenuates sepsis-induced inflammatory responses due to compromised oestrogen deactivation, leading to increased sepsis lethality. In contrast, transgenic overexpression of EST promotes oestrogen deactivation and sensitizes mice to CLP-induced inflammatory response. The induction of EST by sepsis is NF-κB dependent and EST is a NF-κB-target gene. The reciprocal regulation of inflammation and EST may represent a yet-to-be-explored mechanism of endocrine regulation of inflammation, which has an impact on the clinical outcome of sepsis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
524190
5-Chloro-2-(2,4-dichlorophenoxy)phenol, 97%
C12H7Cl3O2
SAB1100404
Anti-RELA (51-65) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300294
Anti-RELA (Ab-254) antibody produced in rabbit, affinity isolated antibody
SAB4300295
Anti-RELA (Ab-276) antibody produced in rabbit, affinity isolated antibody
SAB4300506
Anti-RELA (Ab-311) antibody produced in rabbit, affinity isolated antibody
SAB4300296
Anti-RELA (Ab-435) antibody produced in rabbit, affinity isolated antibody
SAB4300297
Anti-RELA (Ab-468) antibody produced in rabbit, affinity isolated antibody
SAB4300442
Anti-RELA (Ab-505) antibody produced in rabbit, affinity isolated antibody
SAB4300469
Anti-RELA (Ab-529) antibody produced in rabbit, affinity isolated antibody
SAB4300298
Anti-RELA (Ab-536) antibody produced in rabbit, affinity isolated antibody
AV100779
Anti-RELA antibody produced in rabbit, IgG fraction of antiserum
SAB2101980
Anti-RELA antibody produced in rabbit, affinity isolated antibody
SAB2103066
Anti-RELA antibody produced in rabbit, affinity isolated antibody
E8640
E-64d, protease inhibitor
C17H30N2O5
E1024
Estradiol, meets USP testing specifications
C18H24O2
439770
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Cl3Gd
735949
Gadolinium(III) chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl3Gd
721905
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Cl3Gd
721913
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 1 %, Methanol-13C 0.1 % (99 atom % 13C), NMR tube size 5 mm × 8 in.
Cl3Gd
703656
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Cl3Gd
72779
Irgasan, ≥97.0% (HPLC)
C12H7Cl3O2
SAB1404309
Monoclonal Anti-RELA antibody produced in mouse, clone 8G3, ascites fluid
SAB5300389
Monoclonal Anti-RELA antibody produced in mouse, clone 6H7, ascites fluid