EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Etoposide Induces Necrosis Through p53-Mediated Antiapoptosis in Human Kidney Proximal Tubule Cells.
PMID 26259609
Abstract
The p53 protein is an important transcription factor that modulates signaling pathways for both cell death and survival. Its antiapoptotic mechanisms that correlate with necrotic and apoptotic cell death are not well understood. Here, we report that etoposide promotes progression of the DNA damage response as well as necrotic morphological changes including plasma membrane rupture using carbon nanotube-tipped/atomic force microscopy (CNT/AFM) probes in human kidney proximal tubule (HK-2) cells. Inhibition of p53 abrogated cell cycle arrest and led to a decrease in the expression levels of repair proteins that were induced by DNA damage. Mitochondrial biogenesis and cytosolic production of reactive oxygen species were also reduced after p53 inhibition; the latter change induced mitochondrial superoxide accumulation and mitochondrial damage, which triggered the activation of caspase 3. Inhibition of p53 also led to a loss of cell adhesion and converted necrotic cell death to apoptotic cell death, with appreciable cell shrinkage and appearance of apoptotic bodies that were observed using CNT/AFM probes. Thus, our study demonstrated that p53 protects against apoptosis, and leads to etoposide-induced necrosis. These results are expected to aid in the understanding of mechanism of antiapoptosis and its relationship to cell death.