EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
The second virial coefficient as a predictor of protein aggregation propensity: A self-interaction chromatography study.
PMID 26259782
Abstract
The second osmotic virial coefficients (b2) of four proteins - lysozyme, recombinant human lactoferrin, concanavalin A and catalase were measured by self-interaction chromatography (SIC) in solutions of varying salt type, concentration and pH. Protein aggregate sizes based on the initial hydrodynamic radius of the protein solution species present were measured using dynamic light scattering, and the relationship between b2 and protein aggregate size was studied. A linear correlation was established between b2 values and protein aggregate hydrodynamic size for all proteins, and for almost all solution conditions. Aggregate sizes of <∼10nm, indicative of non-aggregated protein systems, were consistently observed to have b2 values >0. The observed b2 trends as a function of solution conditions were very much protein dependent, with notable trends including the existence of attractive interactions (negative b2 values) at low ionic strengths for catalase and concanavalin A, and the highly positive b2 values observed for lactoferrin over a wide range of solution conditions, reflecting lactoferrin's innately high stability. It is concluded that the quantification of protein-protein interactions using SIC based b2 data is a potentially valuable screening tool for predicting protein aggregation propensity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48509
1,2-Dichloroethane, analytical standard
C2H4Cl2
C9322 Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
L7647
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
411000
Ethanolamine, purified by redistillation, ≥99.5%
C2H7NO
398136
Ethanolamine, ACS reagent, ≥99.0%
C2H7NO
ARK2169
Ethanolamine, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for Pharma or biopharmaceutical production
C2H7NO
110167
Ethanolamine, ReagentPlus®, ≥99%
C2H7NO
E9508
Ethanolamine, ≥98%
C2H7NO
E0135
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
C2H7NO
15014
Ethanolamine, ≥99%
C2H7NO
L4040
Lactoferrin human, recombinant, expressed in rice, Partially iron saturated, ≥90% (SDS-PAGE)
L1294 Lactoferrin human, recombinant, expressed in rice, Iron saturated, ≥90% (SDS-PAGE)
P5629
Potassium phosphate tribasic, reagent grade, ≥98%
K3O4P
S7670
Sodium acetate trihydrate, BioXtra, ≥99.0%
C2H3NaO2 · 3H2O
S1304
Sodium acetate trihydrate, meets USP testing specifications
C2H3NaO2 · 3H2O
71188
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
C2H3NaO2 · 3H2O
25022
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
C2H3NaO2 · 3H2O
236500
Sodium acetate trihydrate, ACS reagent, ≥99%
C2H3NaO2 · 3H2O
S8625
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
C2H3NaO2 · 3H2O
42077
Sodium cyanoborohydride, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
NaBH3CN
156159
Sodium cyanoborohydride, reagent grade, 95%
NaBH3CN
296813
Sodium cyanoborohydride solution, 1.0 M in THF
CH3BNNa
296945
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
CH3BNNa