EMAIL THIS PAGE TO A FRIEND
Scientific reports
miR-744 enhances type I interferon signaling pathway by targeting PTP1B in primary human renal mesangial cells.
PMID 26259828
Abstract
Renal mesangial cells (RMCs) constitute a population of cells in glomerular mesangium. Inflammatory cytokines produced by RMCs play a vital role in renal inflammation. miRNAs are key regulators of inflammatory cytokine expression. The abnormal expression of renal miRNAs and the consequent changes in inflammatory signal transduction are closely associated with renal inflammation. However, our knowledge of the functions of renal miRNAs is still limited. In this study, we investigated the role of miR-744 in type I interferon (IFN) signaling pathway in primary human RMCs. We show that overexpression of miR-744 enhances IFN-induced CCL2, CCL5, CXCL10, and IL6 expression specifically in RMCs. We found that the activation of TYK2, STAT1 and STAT3 was significantly enhanced by miR-744. miR-744 also enhanced the activation of non-classical signal components, such as ERK and p38. We then identified PTP1B, a ubiquitously expressed phosphatase, as the target of miR-744 that is responsible for enhancing type I IFN response. Finally, miR-744 expression was induced by type I IFN in RMCs. Collectively, our data indicate that by targeting PTP1B, miR-744 plays a feed-forward role in regulating type I IFN signaling pathway. These findings give us new insights into the functions of renal miRNAs in regulating important signaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300123
Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit, affinity isolated antibody
SAB4504446
Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit, affinity isolated antibody
SAB4300034
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504540
Anti-phospho-STAT3 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2690 Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300033
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4504541
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4300708
Anti-STAT3 (Ab-705) antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406487
Anti-STAT3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001671
Anti-STAT3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
S5933 Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB4502871
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502872
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502873
Anti-STAT3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300392
Anti-TYK2 (Ab-1054) antibody produced in rabbit, affinity isolated antibody
SAB1411977
ANTI-TYK2 antibody produced in mouse, clone 5A4, purified immunoglobulin, buffered aqueous solution
HPA005157
Anti-TYK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503050
Anti-TYK2, C-Terminal antibody produced in rabbit, affinity isolated antibody
145726
DL-Tyrosine, 99%
C9H11NO3
123072
Luminol, 97%
C8H7N3O2
A8511
Luminol, ≥97% (HPLC)
C8H7N3O2
EHU016451 MISSION® esiRNA, esiRNA targeting human PTPN1 (esiRNA1)
EMU061791 MISSION® esiRNA, esiRNA targeting mouse Ptpn1 (esiRNA1)
SIHP0724 MISSION® Human Phosphatase, PTPN1 siRNA1, Nano Scale 250 pmol
SIHP0725 MISSION® Human Phosphatase, PTPN1 siRNA2, Nano Scale 250 pmol