EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Synergistic effects of snail and quercetin on renal cell carcinoma Caki-2 by altering AKT/mTOR/ERK1/2 signaling pathways.
PMID 26261493
Abstract
Renal cell carcinoma has become the most common subtype of kidney cancer, and has the highest propensity to manifest as metastatic disease. Because of lack of knowledge in events that correlated with tumor cell migration and invasion, few therapeutic options are available. Therefore, in current study, we explore the anti-tumoral effect of a potential chemopreventive natural product, quercetin, combined with anti-sense oligo gene therapy (inhibiting Snail gene). We found that either one of them had the remarkable effects in suppressing cell proliferation and migration, inducing cell cycle arrest and apoptosis in a ccRCC cell line, Caki-2 cells. The combination of both means provides even strong suppressive effects toward these ccRCC cells. Our study, for the first time, provides the possibility of using a novel treatment for renal cancer, by combining natural product and gene therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405523
Anti-BSG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA036048
Anti-BSG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502896
Anti-CD147, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4200577
Anti-COX2 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA001335
Anti-COX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200576
Anti-COX2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4502491
Anti-COX2, C-Terminal antibody produced in rabbit, affinity isolated antibody
E4029 Anti-EMMPRIN antibody produced in goat, affinity isolated antibody
E4154 Anti-EMMPRIN antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2501940
Anti-SETD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA042451
Anti-SETD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4100147
Monoclonal Anti-BSG antibody produced in mouse, clone BSG-A114, culture supernatant
SAB4100148
Monoclonal Anti-BSG antibody produced in mouse, clone BSG-B114, culture supernatant
SAB4700021 Monoclonal Anti-CD147 antibody produced in mouse, clone MEM-M6/2, purified immunoglobulin, buffered aqueous solution
SAB4700022 Monoclonal Anti-CD147 antibody produced in mouse, clone MEM-M6/1, purified immunoglobulin, buffered aqueous solution