EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Effects of hydrogen sulfide on high glucose-induced glomerular podocyte injury in mice.
PMID 26261567
Abstract
The aim of this study was to assess the effects of hydrogen sulfide on high glucose-induced mouse podocyte (MPC) injury and the underlying mechanisms. Mouse podocytes were randomly divided into 4 groups, including high glucose (HG), normal glucose (NG), normal glucose + DL-propargylglycine (PPG), and high glucose + NaHS (HG + NaHS) groups for treatment. Then, ZO-2, nephrin, β-catenin, and cystathionine γ-lyase (CSE) protein expression levels were determined by western blot. We found that high glucose significantly reduced nephrin, ZO-2, and CSE expression levels (P<0.05), and overtly elevated β-catenin amounts (P<0.05), in a time-dependent manner. Likewise, PPG at different concentrations in normal glucose resulted in significantly lower CSE, ZO-2, and nephrin levels (P<0.05), and increased β-catenin amounts (P<0.05). Interestingly, significantly increased ZO-2 and nephrin levels, and overtly reduced β-catenin amounts were observed in the HG + NaHS group compared with HG treated cells (P<0.01). Compared with NG treated cells, decreased ZO-2 and nephrin levels and higher β-catenin amounts were obtained in the HG + NaHS group. In conclusion,CSE downregulation contributes to hyperglycemia induced podocyte injury, which is alleviated by exogenous H2S possibly through ZO-2 upregulation and the subsequent suppression of Wnt/β-catenin pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405673
Anti-CTH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV46068
Anti-CTH antibody produced in rabbit, IgG fraction of antiserum
HPA021591
Anti-CTH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA023300
Anti-CTH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2100501
Anti-CTH antibody produced in rabbit, affinity isolated antibody
PRS2265
Anti-Nephrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900278
Anti-NPHS1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500770
Anti-NPHS1 antibody produced in rabbit, affinity isolated antibody
SAB2102435
Anti-TJP2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102436
Anti-TJP2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412778
Anti-TJP2 antibody produced in mouse, purified immunoglobulin, 0.5 mg/mL
HPA001813
Anti-TJP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1403711
Monoclonal Anti-CTH antibody produced in mouse, clone S51, purified immunoglobulin, buffered aqueous solution
SAB1402860
Monoclonal Anti-NPHS1 antibody produced in mouse, clone 3H6, purified immunoglobulin, buffered aqueous solution
SAB1412779
Monoclonal Anti-TJP2 antibody produced in mouse, clone 4D12, purified immunoglobulin, 0.5 mg/mL