EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Klotho gene polymorphisms are related to colorectal cancer susceptibility.
PMID 26261651
Abstract
The purpose of this study was to investigate the relationship of Klotho gene G-395A and C1818T polymorphisms with colorectal cancer (CRC) susceptibility. 125 CRC patients and 125 controls were enrolled in the study. G-395A and C1818T polymorphisms were genotyped with polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP) technology. Haploview software was utilized to conduct linkage disequilibrium and haplotype analysis. Odds ratio (OR) and 95% confidence interval (95% CI) were used to analyze the correlation of genotypes and haplotypes with CRC susceptibility. AA and GA genotypes of G-395A polymorphisms were related with CRC risk (AA: OR = 4.161, 95% CI = 1.437-12.053; GA: OR = 1.958, 95% CI = 1.133-3.385). The frequency of A allele was much higher in case group, compared with controls (31.2% vs.17.6%) and the value of OR AND 95% CI suggested that A allele served as a risk factor for CRC (OR = 2.123, 95% CI = 1.393-3.236). Haplotypes analysis indicated that A-C and A-T haplotypes were significantly associated with risk of CRC (OR = 1.822, 95% CI = 1.124-2.954; OR = 2.877, 95% CI = 1.340-6.176). G-395A polymorphism of Klotho gene could increase the risk of CRC.