EMAIL THIS PAGE TO A FRIEND
BMC research notes
Afrostyrax lepidophyllus extracts exhibit in vitro free radical scavenging, antioxidant potential and protective properties against liver enzymes ion mediated oxidative damage.
PMID 26264271
Abstract
Several studies described the phytochemical constituents of plants in relation with the free radical scavenging property and inhibition of lipid peroxidation. This study investigated the in vitro antioxidant property, and the protective effects of ethanolic and aqueous ethanol extract of the leaves and barks of Afrostyrax lepidophyllus (Huaceae) against ion mediated oxidative damages. Four extracts (ethanol and aqueous-ethanol) from the leaves and barks of A. lepidophyllus were used in this study. The total phenols content, the antiradical and antioxidant properties were determined using standard colorimetric methods. The plant extracts had a significant scavenging potential on the 2,2-diphenyl-1-picrylhydrazyl (DPPH), hydroxyl (OH), nitrite oxide (NO) and 2,2-azinobis (3-ethylbenzthiazoline)-6-sulfonic acid (ABTS) radicals with the IC50 varied between 47 and 200 µg/mL depending on the part of plant and the type of extract. The ethanol extract of A. lepidophyllus bark (GEE) showed the highest polyphenolic (35.33 ± 0.29) and flavonoid (12.00 ± 0.14) content. All the tested extracts demonstrated a high protective potential with the increased of superoxide dismutase, catalase and peroxidase activities. Afrostyrax lepidophyllus extracts exhibited higher antioxidant potential and significant protective potential on liver enzymes.