EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
A Fatality Related to Two Novel Hallucinogenic Compounds: 4-Methoxyphencyclidine and 4-Hydroxy-N-methyl-N-ethyltryptamine.
PMID 26265285
Abstract
In this case report, we present an evaluation of postmortem concentration distribution of the hallucinogenic compound 4-methoxyphencyclidine (4-MeO-PCP) in a fatality principally attributed to this drug. Another hallucinogen, 4-hydroxy-N-methyl-N-ethyltryptamine was also detected, but was not quantitated. A man--who had a history of recent 'strange' behavior--was found deceased, on his bed, in his locked room. Toxicology testing, which initially screened positive for phencyclidine (PCP) by ELISA, subsequently detected and confirmed the two hallucinogens by gas chromatography-mass spectrometry. 4-MeO-PCP concentrations were then quantified by a specific secondary testing technique. The peripheral blood concentration was 8.2 mg/L compared with the central blood concentration of 14 mg/L. The liver concentration was 120 mg/kg, the vitreous was 5.1 mg/L, the urine was 140 mg/L and the gastric contents contained 280 mg. PCP was not detected, but therapeutic concentrations of venlafaxine, olanzapine, lorazepam and hydroxyzine were confirmed. The cause of death was certified due to acute mixed drug intoxication, and the manner of death was certified as accident.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6506
4-Chloro-DL-phenylalanine
C9H10ClNO2