PloS one
A comparison of the biological effects of 125I seeds continuous low-dose-rate radiation and 60Co high-dose-rate gamma radiation on non-small cell lung cancer cells.
PMID 26266801
Abstract
To compare the biological effects of 125I seeds continuous low-dose-rate (CLDR) radiation and 60Co γ-ray high-dose-rate (HDR) radiation on non-small cell lung cancer (NSCLC) cells. A549, H1299 and BEAS-2B cells were exposed to 125I seeds CLDR radiation or 60Co γ-ray HDR radiation. The survival fraction was determined using a colony-forming assay. The cell cycle progression and apoptosis were detected by flow cytometry (FCM). The expression of the apoptosis-related proteins caspase-3, cleaved-caspase-3, PARP, cleaved-PARP, BAX and Bcl-2 were detected by western blot assay. After irradiation with 125I seeds CLDR radiation, there was a lower survival fraction, more pronounced cell cycle arrest (G1 arrest and G2/M arrest in A549 and H1299 cells, respectively) and a higher apoptotic ratio for A549 and H1299 cells than after 60Co γ-ray HDR radiation. Moreover, western blot assays revealed that 125I seeds CLDR radiation remarkably up-regulated the expression of Bax, cleaved-caspase-3 and cleaved-PARP proteins and down-regulated the expression of Bcl-2 proteins in A549 and H1299 cells compared with 60Co γ-ray HDR radiation. However, there was little change in the apoptotic ratio and expression of apoptosis-related proteins in normal BEAS-2B cells receiving the same treatment. 125I seeds CLDR radiation led to remarkable growth inhibition of A549 and H1299 cells compared with 60Co HDR γ-ray radiation; A549 cells were the most sensitive to radiation, followed by H1299 cells. In contrast, normal BEAS-2B cells were relatively radio-resistant. The imbalance of the Bcl-2/Bax ratio and the activation of caspase-3 and PARP proteins might play a key role in the anti-proliferative effects induced by 125I seeds CLDR radiation, although other possibilities have not been excluded and will be investigated in future studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3428 Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500343
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
B8429
Anti-Bax antibody, Mouse monoclonal, clone 6A7, purified from hybridoma cell culture
B8554
Anti-Bax antibody, Mouse monoclonal, clone 2D2, purified from hybridoma cell culture
SAB4502546
Anti-BAX, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502547
Anti-BAX, C-Terminal antibody produced in rabbit, affinity isolated antibody
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B9804 Anti-Bcl-2 antibody, Mouse monoclonal, clone 10C4, purified from hybridoma cell culture
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
SAB2500154
Anti-BCL2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2101602
Anti-NKX3-1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101603
Anti-NKX3-1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412056
ANTI-NKX3-1 antibody produced in mouse, clone 3C1, purified immunoglobulin, buffered aqueous solution
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3700855 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution