PloS one
Cortical Granule Exocytosis Is Mediated by Alpha-SNAP and N-Ethilmaleimide Sensitive Factor in Mouse Oocytes.
PMID 26267363
Abstract
Cortical granule exocytosis (CGE), also known as cortical reaction, is a calcium- regulated secretion that represents a membrane fusion process during meiotic cell division of oocytes. The molecular mechanism of membrane fusion during CGE is still poorly understood and is thought to be mediated by the SNARE pathway; nevertheless, it is unkown if SNAP (acronym for soluble NSF attachment protein) and NSF (acronym for N-ethilmaleimide sensitive factor), two key proteins in the SNARE pathway, mediate CGE in any oocyte model. In this paper, we documented the gene expression of α-SNAP, γ-SNAP and NSF in mouse oocytes. Western blot analysis showed that the expression of these proteins maintains a similar level during oocyte maturation and early activation. Their localization was mainly observed at the cortical region of metaphase II oocytes, which is enriched in cortical granules. To evaluate the function of these proteins in CGE we set up a functional assay based on the quantification of cortical granules metaphase II oocytes activated parthenogenetically with strontium. Endogenous α-SNAP and NSF proteins were perturbed by microinjection of recombinant proteins or antibodies prior to CGE activation. The microinjection of wild type α-SNAP and the negative mutant of α-SNAP L294A in metaphase II oocytes inhibited CGE stimulated by strontium. NEM, an irreversibly inhibitor of NSF, and the microinjection of the negative mutant NSF D1EQ inhibited cortical reaction. The microinjection of anti-α-SNAP and anti-NSF antibodies was able to abolish CGE in activated metaphase II oocytes. The microinjection of anti-γ SNAP antibody had no effect on CGE. Our findings indicate, for the first time in any oocyte model, that α-SNAP, γ-SNAP, and NSF are expressed in mouse oocytes. We demonstrate that α-SNAP and NSF have an active role in CGE and propose a working model.