EMAIL THIS PAGE TO A FRIEND
Nature communications
A hepatic amino acid/mTOR/S6K-dependent signalling pathway modulates systemic lipid metabolism via neuronal signals.
PMID 26268630
Abstract
Metabolism is coordinated among tissues and organs via neuronal signals. Levels of circulating amino acids (AAs), which are elevated in obesity, activate the intracellular target of rapamycin complex-1 (mTORC1)/S6kinase (S6K) pathway in the liver. Here we demonstrate that hepatic AA/mTORC1/S6K signalling modulates systemic lipid metabolism via a mechanism involving neuronal inter-tissue communication. Hepatic expression of an AA transporter, SNAT2, activates the mTORC1/S6K pathway, and markedly elevates serum triglycerides (TGs), while downregulating adipose lipoprotein lipase (LPL). Hepatic Rheb or active-S6K expression have similar metabolic effects, whereas hepatic expression of dominant-negative-S6K inhibits TG elevation in SNAT2 mice. Denervation, pharmacological deafferentation and β-blocker administration suppress obesity-related hypertriglyceridemia with adipose LPL upregulation, suggesting that signals are transduced between liver and adipose tissue via a neuronal pathway consisting of afferent vagal and efferent sympathetic nerves. Thus, the neuronal mechanism uncovered here serves to coordinate amino acid and lipid levels and contributes to the development of obesity-related hypertriglyceridemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB152
Anti-Tyrosine Hydroxylase Antibody, 100 µL, Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, IF, IH, IH(P), IP and WB.
SAB2101327
Anti-LCP1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101328
Anti-LCP1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2701641
Anti-lcp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701743
Anti-lcp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA019493
Anti-LCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406091
Anti-LPL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401231
Anti-LPL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2700761
Anti-LPL antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701143
Anti-LPL antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701144
Anti-LPL antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33244
Anti-RORC (AB1) antibody produced in rabbit, affinity isolated antibody
AV45622
Anti-RORC (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2501680
Anti-RORC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102985
Anti-RORC antibody produced in rabbit, affinity isolated antibody
SAB2105313
Anti-RORC, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA035180
Anti-SLC38A2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502246
Anti-SLC38A2 antibody produced in rabbit, affinity isolated antibody
SAB2104027
Anti-SLC38A2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
PRS3485
Anti-TOR antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5300056
Monoclonal Anti-LPL antibody produced in mouse, clone 2C5, ascites fluid
SAB1402341
Monoclonal Anti-RORC, (C-terminal) antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution