EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Simultaneous Determination of 10 Ultratrace Elements in Infant Formula, Adult Nutritionals, and Milk Products by ICP/MS After Pressure Digestion: Single-Laboratory Validation.
PMID 26268978
Abstract
A single-laboratory validation (SLV) is presented for the simultaneous determination of 10 ultratrace elements (UTEs) including aluminum (Al), arsenic (As), cadmium (Cd), cobalt (Co), chromium (Cr), mercury (Hg), molybdenum (Mo), lead (Pb), selenium (Se), and tin (Sn) in infant formulas, adult nutritionals, and milk based products by inductively coupled plasma (ICP)/MS after acidic pressure digestion. This robust and routine multielemental method is based on several official methods with modifications of sample preparation using either microwave digestion or high pressure ashing and of analytical conditions using ICP/MS with collision cell technology. This SLV fulfills AOAC method performance criteria in terms of linearity, specificity, sensitivity, precision, and accuracy and fully answers most international regulation limits for trace contaminants and/or recommended nutrient levels established for 10 UTEs in targeted matrixes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
461210
Cerium, chips
Ce
261041
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Ce
263001
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Ce
GF97217220
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Ce
GF49222582
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Ce
GF61818363
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF27295724
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF97378677
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Ce
GF36157394
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Ce
GF90687695
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ce
GF39030353
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Ce
GF04444095
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Ce
GF55512244
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Ce
GF02435554
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Ce
GF35961655
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ce
GF49695047
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Ce
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y