EMAIL THIS PAGE TO A FRIEND
Pflugers Archiv : European journal of physiology
UCP2 modulates single-channel properties of a MCU-dependent Ca(2+) inward current in mitochondria.
PMID 26275882
Abstract
The mitochondrial Ca(2+) uniporter is a highly Ca(2+)-selective protein complex that consists of the pore-forming mitochondrial Ca(2+) uniporter protein (MCU), the scaffolding essential MCU regulator (EMRE), and mitochondrial calcium uptake 1 and 2 (MICU1/2), which negatively regulate mitochondrial Ca(2+) uptake. We have previously reported that uncoupling proteins 2 and 3 (UCP2/3) are also engaged in the activity of mitochondrial Ca(2+) uptake under certain conditions, while the mechanism by which UCP2/3 facilitates mitochondrial Ca(2+) uniport remains elusive. This work was designed to investigate the impact of UCP2 on the three distinct mitochondrial Ca(2+) currents found in mitoplasts isolated from HeLa cells, the intermediate- (i-), burst- (b-) and extra-large (xl-) mitochondrial/mitoplast Ca(2+) currents (MCC). Using the patch clamp technique on mitoplasts from cells with reduced MCU and EMRE unveiled a very high affinity of MCU for xl-MCC that succeeds that for i-MCC, indicating the coexistence of at least two MCU/EMRE-dependent Ca(2+) currents. The manipulation of the expression level of UCP2 by either siRNA-mediated knockdown or overexpression changed exclusively the open probability (NPo) of xl-MCC by approx. 38% decrease or nearly a 3-fold increase, respectively. These findings confirm a regulatory role of UCP2 in mitochondrial Ca(2+) uptake and identify UCP2 as a selective modulator of just one distinct MCU/EMRE-dependent mitochondrial Ca(2+) inward current.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8874
CGP-37157, ≥98% (HPLC), powder
C15H11Cl2NOS