PloS one
Combined Salvianolic Acid B and Ginsenoside Rg1 Exerts Cardioprotection against Ischemia/Reperfusion Injury in Rats.
PMID 26280455
Abstract
Lack of pharmacological strategies in clinics restricts the patient prognosis with myocardial ischemia/reperfusion (I/R) injury. The aim of this study was to evaluate the cardioprotection of combined salvianolic acid B (SalB) and ginsenoside Rg1 (Rg1) against myocardial I/R injury and further investigate the underlying mechanism. I/R injury was induced by coronary artery ligation for Wistar male rats and hypoxia/reoxygenation injury was induced on H9c2 cells. Firstly, the best ratio between SalB and Rg1was set as 2:5 based on their effects on heart function detected by hemodynamic measurement. Then SalB-Rg1 (2:5) was found to maintain mitochondrial membrane potential and resist apoptosis and necrosis in H9c2 cell with hypoxia/reoxygenation injury. Companying with same dose of SalB or Rg1 only, SalB-Rg1 showed more significant effects on down-regulation of myocardial infarct size, maintenance of myocardium structure, improvement on cardiac function, decrease of cytokine secretion including TNF-α, IL-1β, RANTES and sVCAM-1. Finally, the SalB-Rg1 improved the viability of cardiac myocytes other than cardiac fibroblasts in rats with I/R injury using flow cytometry. Our results revealed that SalB-Rg1 was a promising strategy to prevent myocardial I/R injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8383
Chloral hydrate
C2H3Cl3O2
15307
Chloral hydrate, meets analytical specification of Ph. Eur., BP, USP, 99.5-101%
C2H3Cl3O2
M2643
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
MgSO4
203726
Magnesium sulfate, ≥99.99% trace metals basis
MgSO4
M3409
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
MgSO4
83266
Magnesium sulfate solution, BioUltra, for molecular biology
MgSO4
P5655
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
H2KO4P
RES20760-A7
Potassium phosphate monobasic, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H2KO4P
P8416
Potassium phosphate monobasic, plant cell culture tested, ≥99%
H2KO4P
P9791
Potassium phosphate monobasic, for molecular biology, ≥98.0%
H2KO4P
229806
Potassium phosphate monobasic, 99.99% trace metals basis
H2KO4P
P5379
Potassium phosphate monobasic, ReagentPlus®
H2KO4P
60218
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
H2KO4P
S5761
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
CHNaO3
S6297
Sodium bicarbonate, BioXtra, 99.5-100.5%
CHNaO3
S4019
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
CHNaO3
487031
Sodium bicarbonate-12C, 99.9 atom % 12C
12CHNaO3