EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Impact of Toxoplasma gondii on Dendritic Cell Subset Function in the Intestinal Mucosa.
PMID 26283477
Abstract
The function of mucosal dendritic cell (DC) subsets in immunity and inflammation is not well understood. In this study, we define four DC subsets present within the lamina propria and mesenteric lymph node compartments based on expression of CD103 and CD11b. Using IL-12p40 YFP (Yet40) reporter mice, we show that CD103(+)CD11b(-) mucosal DCs are primary in vivo sources of IL-12p40; we also identified CD103(-)CD11b(-) mucosal DCs as a novel population producing this cytokine. Infection was preferentially found in CD11b(+) DCs that were negative for CD103. Lamina propria DCs containing parasites were negative for IL-12p40. Instead, production of the cytokine was strictly a property of noninfected cells. We also show that vitamin A metabolism, as measured by ALDH activity, was preferentially found in CD103(+)CD11b(+) DC and was strongly downregulated in all mucosal DC subsets during infection. Finally, overall apoptosis of lamina propria DC subsets was increased during infection. Combined, these results highlight the ability of intestinal Toxoplasma infection to alter mucosal DC activity at both the whole population level and at the level of individual subsets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA026761
Anti-EPCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502031
Anti-IRF4 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1411959
ANTI-IRF4 antibody produced in mouse, clone 3A10, purified immunoglobulin, buffered aqueous solution
SAB1412642
ANTI-IRF4 antibody produced in mouse, clone 3C4, purified immunoglobulin, buffered aqueous solution
HPA002038
Anti-IRF4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV38154
Anti-IRF4 antibody produced in rabbit, affinity isolated antibody
AV32669
Anti-IRF4 antibody produced in rabbit, IgG fraction of antiserum
SAB2101176
Anti-IRF4 antibody produced in rabbit, affinity isolated antibody
HPA002698
Anti-IRF4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501566
Anti-IRF4 antibody produced in rabbit, affinity isolated antibody
HPA002267
Anti-IRF8 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100841
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
HPA002531
Anti-IRF8 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103913
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
SAB2107842
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
SAB1400156
Anti-TACSTD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1400157
Anti-TACSTD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB4700423 Monoclonal Anti-CD326 antibody produced in mouse, clone VU-1D9, purified immunoglobulin, buffered aqueous solution
WH0003662M2
Monoclonal Anti-IRF4 antibody produced in mouse, clone 2F2, purified immunoglobulin, buffered aqueous solution
SAB1403990
Monoclonal Anti-IRF4, (C-terminal) antibody produced in mouse, clone 2D1, purified immunoglobulin, buffered aqueous solution