EMAIL THIS PAGE TO A FRIEND
Scientific reports
Virulence from vesicles: Novel mechanisms of host cell injury by Escherichia coli O104:H4 outbreak strain.
PMID 26283502
Abstract
The highly virulent Escherichia coli O104:H4 that caused the large 2011 outbreak of diarrhoea and haemolytic uraemic syndrome secretes blended virulence factors of enterohaemorrhagic and enteroaggregative E. coli, but their secretion pathways are unknown. We demonstrate that the outbreak strain releases a cocktail of virulence factors via outer membrane vesicles (OMVs) shed during growth. The OMVs contain Shiga toxin (Stx) 2a, the major virulence factor of the strain, Shigella enterotoxin 1, H4 flagellin, and O104 lipopolysaccharide. The OMVs bind to and are internalised by human intestinal epithelial cells via dynamin-dependent and Stx2a-independent endocytosis, deliver the OMV-associated virulence factors intracellularly and induce caspase-9-mediated apoptosis and interleukin-8 secretion. Stx2a is the key OMV component responsible for the cytotoxicity, whereas flagellin and lipopolysaccharide are the major interleukin-8 inducers. The OMVs represent novel ways for the E. coli O104:H4 outbreak strain to deliver pathogenic cargoes and injure host cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409834
Anti-CCDC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019049
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019051
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2107667
Anti-CCDC6 antibody produced in rabbit, affinity isolated antibody
SAB1411455
Anti-CCDC6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410334
Anti-EMP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1300570
Anti-HRH4 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2701955
Anti-HRH4 (N-terminal) antibody produced in rabbit
HPA035009
Anti-HRH4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502598
Anti-HRH4 antibody produced in rabbit, affinity isolated antibody
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306778
ANTI-ICOS(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2101177
Anti-IRF6 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101178
Anti-IRF6 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2101179
Anti-IRF6 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2500545
Anti-IRF6 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406044
Anti-IRF6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV43608
Anti-IRF6 antibody produced in rabbit, IgG fraction of antiserum
SAB2102995
Anti-IRF6 antibody produced in rabbit, affinity isolated antibody
SAB1400467
Anti-LY96 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1410439
Anti-LY96 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
PRS3289
Anti-MD-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500891
Anti-MD-2 antibody produced in rabbit, clone 9F1B1, affinity isolated antibody
SAB1300786 Anti-MD2 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300785 Anti-MD2 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2102105
Anti-SELS antibody produced in rabbit, affinity isolated antibody
SAB1411081
Anti-SELS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407701
Anti-SELS, (N-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015573
Anti-TLR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500374
Anti-TLR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution