EMAIL THIS PAGE TO A FRIEND
Oncotarget
Simultaneous inhibition of multiple oncogenic miRNAs by a multi-potent microRNA sponge.
PMID 26284487
Abstract
The roles of oncogenic miRNAs are widely recognized in many cancers. Inhibition of single miRNA using antagomiR can efficiently knock-down a specific miRNA. However, the effect is transient and often results in subtle phenotype, as there are other miRNAs contribute to tumorigenesis. Here we report a multi-potent miRNA sponge inhibiting multiple miRNAs simultaneously. As a model system, we targeted miR-21, miR-155 and miR-221/222, known as oncogenic miRNAs in multiple tumors including breast and pancreatic cancers. To achieve efficient knockdown, we generated perfect and bulged-matched miRNA binding sites (MBS) and introduced multiple copies of MBS, ranging from one to five, in the multi-potent miRNA sponge. Luciferase reporter assay showed the multi-potent miRNA sponge efficiently inhibited 4 miRNAs in breast and pancreatic cancer cells. Furthermore, a stable and inducible version of the multi-potent miRNA sponge cell line showed the miRNA sponge efficiently reduces the level of 4 target miRNAs and increase target protein level of these oncogenic miRNAs. Finally, we showed the miRNA sponge sensitize cells to cancer drug and attenuate cell migratory activity. Altogether, our study demonstrates the multi-potent miRNA sponge is a useful tool to examine the functional impact of simultaneous inhibition of multiple miRNAs and proposes a therapeutic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401144
Anti-FPR2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA029154 Anti-FPR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43141
Anti-MKRN1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV43142
Anti-MKRN1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501704
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501717
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102989
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody
SAB2700796
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700798
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700971
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048684
Anti-TRIM27 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1403837
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2G8, purified immunoglobulin, buffered aqueous solution
SAB1402202
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2D8, purified immunoglobulin, buffered aqueous solution
WH0023608M1
Monoclonal Anti-MKRN1 antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution