EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Cardiac atrial appendage stem cells engraft and differentiate into cardiomyocytes in vivo: A new tool for cardiac repair after MI.
PMID 26285182
Abstract
This study assessed whether autologous transplantation of cardiac atrial appendage stem cells (CASCs) preserves cardiac function after myocardial infarction (MI) in a minipig model. CASCs were isolated from right atrial appendages of Göttingen minipigs based on high aldehyde dehydrogenase activity and expanded. MI was induced by a 2h snare ligation of the left anterior descending coronary artery. Upon reperfusion, CASCs were intramyocardially injected under NOGA guidance (MI-CASC, n=10). Non-transplanted pigs (MI, n=8) received sham treatment. 3D electromechanical mapping (EMM) and cardiac MRI were performed to assess left ventricular (LV) function. MI pigs developed LV dilatation at 2 months (2M), while in the MI-CASC group volumes remained stable. Global LV ejection fraction decreased by 16 ± 8% in MI animals vs 3 ± 10% in MI-CASC animals and regional wall thickening in border areas was better preserved in the MI-CASC group. EMM showed decreased viability and wall motion in the LV for both groups POST-MI, whereas at 2M these parameters only improved in the MI-CASC. Substantial cell retention was accompanied by cardiomyogenic differentiation in 98±1% of the transplanted CASCs, which functionally integrated. Second harmonic generation microscopy confirmed the formation of mature sarcomeres in transplanted CASCs. Absence of cardiac arrhythmias indicated the safety of CASC transplantation. CASCs preserve cardiac function by extensive engraftment and cardiomyogenic differentiation. Our data indicate the enormous potential of CASCs in myocardial repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501173
Anti-Connexin 43 antibody produced in rabbit, affinity isolated antibody
SAB4501174
Anti-Connexin 43, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501175
Anti-Connexin 43, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300504
Anti-GJA1 (Ab-367) antibody produced in rabbit, affinity isolated antibody
SAB2501246
Anti-GJA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV36598
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
SAB2900237
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100918
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
HPA019763
Anti-MYL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039262
Anti-MYL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406590
Anti-VWF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001815
Anti-VWF antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002082
Anti-VWF antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408640
Anti-VWF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0002697M1
Monoclonal Anti-GJA1 antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution
WH0004633M1
Monoclonal Anti-MYL2 antibody produced in mouse, clone 3B9-B4, purified immunoglobulin, buffered aqueous solution
SAB5300243
Monoclonal Anti-MYL2 antibody produced in mouse, clone 7C9, ascites fluid
SAB1401660 Monoclonal Anti-MYLPF antibody produced in mouse, clone 3H3, purified immunoglobulin, buffered aqueous solution
AMAB90928
Monoclonal Anti-VWF antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1950, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90931
Monoclonal Anti-VWF antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1957, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402960
Monoclonal Anti-VWF antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution