EMAIL THIS PAGE TO A FRIEND
Scientific reports
Myosin Binding Protein-C Slow Phosphorylation is Altered in Duchenne Dystrophy and Arthrogryposis Myopathy in Fast-Twitch Skeletal Muscles.
PMID 26287277
Abstract
Myosin Binding Protein-C slow (sMyBP-C), encoded by MYBPC1, comprises a family of regulatory proteins of skeletal muscles that are phosphorylated by PKA and PKC. MYBPC1 missense mutations are linked to the development of Distal Arthrogryposis-1 (DA-1). Although structure-function details for this myopathy are evolving, function is undoubtedly driven by sequence variations and post-translational modifications in sMyBP-C. Herein, we examined the phosphorylation profile of sMyBP-C in mouse and human fast-twitch skeletal muscles. We used Flexor Digitorum Brevis (FDB) isolated from young (~2-months old) and old (~14-months old) wild type and mdx mice, and human Abductor Hallucis (AH) and gastrocnemious muscles carrying the DA-1 mutations. Our results indicate both constitutive and differential phosphorylation of sMyBP-C in aged and diseased muscles. We report a 7-35% reduction in the phosphorylation levels of select sites in old wild type and young or old mdx FDB mouse muscles, compared to young wild type tissue. Similarly, we observe a 30-70% decrease in the phosphorylation levels of all PKA and PKC phospho-sites in the DA-1 AH, but not gastrocnemius, muscle. Overall, our studies show that the phosphorylation pattern of sMyBP-C is differentially regulated in response to age and disease, suggesting that phosphorylation plays important roles in these processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501461
Anti-HSP90A, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2101093
Anti-HSP90AA1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2900334
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107614
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB2107615
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB4300541
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
SAB2701832
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701806
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09016
Anti-HSP90AB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2107344
Anti-HSP90AB1 antibody produced in rabbit, affinity isolated antibody
SAB4501462
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
SAB4501463
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
AV09018 Anti-HSPCB antibody produced in rabbit, IgG fraction of antiserum
SAB1305467
ANTI-HSPCB(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21241 Anti-HSPN antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB5300449
Monoclonal Anti-HSP90AA1 antibody produced in mouse, clone 5G5, purified immunoglobulin, buffered aqueous solution
SAB5300085 Monoclonal Anti-HSP90AB1 antibody produced in mouse, clone 5G4, ascites fluid
SAB5300086 Monoclonal Anti-HSP90AB1 antibody produced in mouse, clone 1D9, ascites fluid