EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Lipoxin and resolvin biosynthesis is dependent on 5-lipoxygenase activating protein.
PMID 26289316
Abstract
Resolution of acute inflammation is an active process coordinated by proresolving lipid mediators (SPMs) such as lipoxins (LXs) and resolvins (Rvs), which are formed by the concerted action of 2 lipoxygenases (LOs). Because the exact molecular mechanisms of SPM biosynthesis are not completely understood, we aimed to investigate LX and D-type Rv formation in human leukocytes and HEK293T cells overexpressing leukotriene (LT) pathway enzymes. Activity assays in precursor (15-hydroxyeicosatetraenoic acids, 17-HDoHE)-treated granulocytes [polymorphonuclear leukocytes (PMNLs)] showed a strict dependence of LXA4/RvD1 biosynthesis on cell integrity, and incubation with recombinant human 5-LO did not lead to LX or Rv formation. Pharmacologic inhibition of 5-LO activating protein (FLAP) by MK-886 inhibited LXA4/RvD1 biosynthesis in precursor-treated PMNLs (drug concentration causing 50% inhibition ∼ 0.3/0.2 µM), as did knockdown of the enzyme in MM6 cells, and precursor-treated HEK293T overexpressing 5-LO produced high amounts of LXA4 only in the presence of FLAP. In addition, inhibition of cytosolic phospholipase A2α (cPLA2α) interfered with LXA4/RvD1 formation from exogenous precursors in PMNLs. Furthermore, inhibition of the LT synthases LTA4 hydrolase and LTC4 synthase in PMNL/platelet coincubations augmented LXA4 levels. These findings show that several enzymes known to be involved in the biosynthesis of proinflammatory LTs, such as FLAP and cPLA2α, also contribute to LX and Rv formation.