EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Evidence of the activation of unfolded protein response in granulosa and cumulus cells during follicular growth and maturation.
PMID 26291812
Abstract
The objective of the present study is to investigate whether unfolded protein response (UPR), activated by endoplasmic reticulum (ER) stress, in granulosa cells (GC) and cumulus cells (CC) is involved in the process of follicular growth and maturation. First, to examine the presence of UPR in growing follicles, the expression of spliced form of X-box-binding protein 1 (XBP1(S)) and heat shock 70 kDa protein 5 (HSPA5) mRNA, typical UPR genes, in mice ovaries were examined by in situ hybridization. GC of later stage than large secondary follicles expressed both XBP1(S) and HSPA5 mRNA, which was accompanied with the activation of ER stress sensor proteins, inositol-requiring enzyme 1 (IRE1) and double-stranded RNA-activated protein kinase-like ER kinase (PERK), confirmed by immunohistochemistry. Next, to examine the association between the fertilization capacity of oocytes and the expression levels of UPR genes in surrounding CC, human CC were collected from patients undergoing ICSI. The expression levels of XBP1(S) mRNA, quantified by RT-PCR, in CC enclosing human oocytes achieving fertilization were two-fold higher, than those in CC enclosing oocytes without fertilization capacity. In conclusion, UPR is activated during follicular growth and suggested to be involved in the process of follicular growth and maturation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA015737 Anti-EIF2AK3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2900917
Anti-EIF2AK3 antibody produced in rabbit, affinity isolated antibody
SAB1412027
ANTI-ERN1 antibody produced in mouse, clone 1F12, purified immunoglobulin, buffered aqueous solution
SAB1412028
ANTI-ERN1 antibody produced in mouse, clone 5F3, purified immunoglobulin, buffered aqueous solution
SAB1412029
ANTI-ERN1 antibody produced in mouse, clone 6G11, purified immunoglobulin, buffered aqueous solution
SAB1412341
ANTI-ERN1 antibody produced in mouse, clone 4E1, purified immunoglobulin, buffered aqueous solution
SAB1412645
ANTI-ERN1 antibody produced in mouse, clone 4D12, purified immunoglobulin, buffered aqueous solution
HPA027730 Anti-ERN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3655
Anti-IRE1P (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3657
Anti-IRE1P (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3659
Anti-IRE1P (ab3) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P0074
Anti-PERK (C-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306427
ANTI-PERK (N-TERM Q163) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
P0073
Anti-PERK (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB5300109
Monoclonal Anti-EIF2AK3 antibody produced in mouse, clone 5G5, ascites fluid
SAB1412781
Monoclonal Anti-EIF2AK3 antibody produced in mouse, clone 7B10, purified immunoglobulin, 1 mg/mL
WH0002081M2
Monoclonal Anti-ERN1 antibody produced in mouse, clone 8D12, purified immunoglobulin, buffered aqueous solution
SAB1403792
Monoclonal Anti-ERN1 antibody produced in mouse, clone 3D1, purified immunoglobulin, buffered aqueous solution
SAB5300474 Monoclonal Anti-ERN1 antibody produced in mouse, clone 9F2, ascites fluid