PloS one
VSX2 and ASCL1 Are Indicators of Neurogenic Competence in Human Retinal Progenitor Cultures.
PMID 26292211
Abstract
Three dimensional (3D) culture techniques are frequently used for CNS tissue modeling and organoid production, including generation of retina-like tissues. A proposed advantage of these 3D systems is their potential to more closely approximate in vivo cellular microenvironments, which could translate into improved manufacture and/or maintenance of neuronal populations. Visual System Homeobox 2 (VSX2) labels all multipotent retinal progenitor cells (RPCs) and is known to play important roles in retinal development. In contrast, the proneural transcription factor Acheate scute-like 1 (ASCL1) is expressed transiently in a subset of RPCs, but is required for the production of most retinal neurons. Therefore, we asked whether the presence of VSX2 and ASCL1 could gauge neurogenic potential in 3D retinal cultures derived from human prenatal tissue or ES cells (hESCs). Short term prenatal 3D retinal cultures displayed multiple characteristics of human RPCs (hRPCs) found in situ, including robust expression of VSX2. Upon initiation of hRPC differentiation, there was a small increase in co-labeling of VSX2+ cells with ASCL1, along with a modest increase in the number of PKCα+ neurons. However, 3D prenatal retinal cultures lost expression of VSX2 and ASCL1 over time while concurrently becoming refractory to neuronal differentiation. Conversely, 3D optic vesicles derived from hESCs (hESC-OVs) maintained a robust VSX2+ hRPC population that could spontaneously co-express ASCL1 and generate photoreceptors and other retinal neurons for an extended period of time. These results show that VSX2 and ASCL1 can serve as markers for neurogenic potential in cultured hRPCs. Furthermore, unlike hESC-OVs, maintenance of 3D structure does not independently convey an advantage in the culture of prenatal hRPCs, further illustrating differences in the survival and differentiation requirements of hRPCs extracted from native tissue vs. those generated entirely in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1412009
ANTI-ASCL1 antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
SAB1412150
ANTI-ASCL1 antibody produced in mouse, clone 3D3, purified immunoglobulin, buffered aqueous solution
AV32355
Anti-ASCL1 antibody produced in rabbit, IgG fraction of antiserum
SAB2103889
Anti-ASCL1 antibody produced in rabbit, affinity isolated antibody
AV31974
Anti-CHX10 antibody produced in rabbit, affinity isolated antibody
SAB1300371
Anti-PKCγ (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104731
Anti-PRKCG antibody produced in rabbit, affinity isolated antibody
SAB2104573
Anti-PRKCG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV38232
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104660
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1409219
Anti-VSX2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003436
Anti-VSX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000429M1
Monoclonal Anti-ASCL1 antibody produced in mouse, clone 7E11, purified immunoglobulin, buffered aqueous solution
SAB1403577
Monoclonal Anti-ASCL1, (C-terminal) antibody produced in mouse, clone 2D9, purified immunoglobulin, buffered aqueous solution
S1451
Monoclonal Anti-Sox2 antibody produced in mouse, ~1.0 mg/mL, clone SOX2-6, purified immunoglobulin, buffered aqueous solution
SAB5300177
Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10, ascites fluid
SAB5300369 Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10C9, ascites fluid