EMAIL THIS PAGE TO A FRIEND
Arthritis research & therapy
Plexin-D1/Semaphorin 3E pathway may contribute to dysregulation of vascular tone control and defective angiogenesis in systemic sclerosis.
PMID 26292963
Abstract
The vascular and nervous systems have several anatomic and molecular mechanism similarities. Emerging evidence suggests that proteins involved in transmitting axonal guidance cues, including members of class III semaphorin (Sema3) family, play a critical role in blood vessel guidance during physiological and pathological vascular development. Sema3E is a natural antiangiogenic molecule that causes filopodial retraction in endothelial cells, inhibiting cell adhesion by disrupting integrin-mediated adhesive structures. The aim of the present study was to investigate whether in systemic sclerosis (SSc) Plexin-D1/Sema3E axis could be involved in the dysregulation of vascular tone control and angiogenesis. Sema3E levels were measured by quantitative colorimetric sandwich ELISA in serum samples from 48 SSc patients, 45 subjects with primary Raynaud's phenomenon (pRP) and 48 age-matched and sex-matched healthy controls. Immunofluorescence staining on skin sections from 14 SSc patients and 12 healthy subjects was performed to evaluate Sema3E and Plexin-D1 expression. Western blotting was used to assess Plexin-D1/Sema3E axis in human SSc and healthy dermal microvascular endothelial cells (SSc-MVECs and H-MVECs, respectively) at basal condition and after stimulation with recombinant human vascular endothelial growth factor (VEGF), SSc and healthy sera. Capillary morphogenesis on Matrigel was performed on H-MVECs treated with healthy, pRP or SSc sera in the presence of Sema3E and Plexin-D1 soluble peptides. Serum Sema3E levels were significantly higher both in pRP subjects and SSc patients than in controls. In SSc, Sema3E levels were significantly increased in patients with early nailfold videocapillaroscopy (NVC) pattern compared to active/late patterns and pRP, and in patients without digital ulcers versus those with ulcers. In SSc skin, Sema3E expression was strongly increased in the microvascular endothelium. Cultured SSc-MVECs showed higher levels of phosphorylated Plexin-D1 and Sema3E expression than H-MVECs, and stimulation with SSc sera increased phosphorylated Plexin-D1 and Sema3E in H-MVECs. The addition of Sema3E-binding Plexin-D1 soluble peptide significantly attenuated the antiangiogenic effect of SSc sera on H-MVECs. Our findings suggest that Plexin-D1/Sema3E axis is triggered in SSc endothelium and may have a role in the dysregulation of angiogenesis and vascular tone control by inducing neuro-vascular mechanism alterations clinically evident in particular in the early disease phases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA021289
Anti-BBS9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408674
Anti-BBS9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306529
ANTI-CD31(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1304992 ANTI-DIO1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3700280 Anti-goat IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700328 Anti-Goat IgG (H+L), F(ab) fragment- antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB1407254
Anti-LMOD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101354
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB4200200
Anti-LMOD1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA028325
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028435
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030097
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB2107079
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB4502167
Anti-PECAM-1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406243
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408616
Anti-PECAM1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1305785
ANTI-PTHB1(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3700932 Anti-Rabbit IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700976 Anti-Rabbit IgG (H+L), F(ab) fragment antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
B4429
BIS-TRIS, BioPerformance Certified, cell culture tested, suitable for insect cell culture, ≥98.0%
C8H19NO5
B9754
BIS-TRIS, ≥98.0% (titration)
C8H19NO5
RES1161B-A7
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
C8H19NO5
PHG0004
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H19NO5
B7535
BIS-TRIS, BioXtra, ≥98.0% (titration)
C8H19NO5
14879
BIS-TRIS, BioUltra, ≥99.0% (NT)
C8H19NO5
SAB4700463
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
SAB1402173 Monoclonal Anti-DIO1 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
SAB4200496
Monoclonal Anti-LMOD1 antibody produced in mouse, ~1.0 mg/mL, clone LMOD1-12, purified immunoglobulin
SAB1401605
Monoclonal Anti-LMOD1 antibody produced in mouse, clone 3B8, purified immunoglobulin, buffered aqueous solution
WH0005175M1
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution