EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
An Nfic-hedgehog signaling cascade regulates tooth root development.
PMID 26293299
Abstract
Coordination between the Hertwig's epithelial root sheath (HERS) and apical papilla (AP) is crucial for proper tooth root development. The hedgehog (Hh) signaling pathway and Nfic are both involved in tooth root development; however, their relationship has yet to be elucidated. Here, we establish a timecourse of mouse molar root development by histological staining of sections, and we demonstrate that Hh signaling is active before and during root development in the AP and HERS using Gli1 reporter mice. The proper pattern of Hh signaling activity in the AP is crucial for the proliferation of dental mesenchymal cells, because either inhibition with Hh inhibitors or constitutive activation of Hh signaling activity in transgenic mice leads to decreased proliferation in the AP and shorter roots. Moreover, Hh activity is elevated in Nfic(-/-) mice, a root defect model, whereas RNA sequencing and in situ hybridization show that the Hh attenuator Hhip is downregulated. ChIP and RNAscope analyses suggest that Nfic binds to the promoter region of Hhip. Treatment of Nfic(-/-) mice with Hh inhibitor partially restores cell proliferation, AP growth and root development. Taken together, our results demonstrate that an Nfic-Hhip-Hh signaling pathway is crucial for apical papilla growth and proper root formation. This discovery provides insight into the molecular mechanisms regulating tooth root development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40725
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
C5H10N2O
40727
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
C5H10N2O
193453
1,3-Dimethyl-2-imidazolidinone, reagent grade
C5H10N2O
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101580
Anti-NFIC antibody produced in rabbit, affinity isolated antibody
SAB2103065
Anti-NFIC antibody produced in rabbit, affinity isolated antibody
SAB1410837
Anti-NFIC antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C8267 Corn oil, delivery vehicle for fat-soluble compounds
WH0000836M2 Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
WH0004782M1
Monoclonal Anti-NFIC antibody produced in mouse, clone 1C12-2A2, purified immunoglobulin, buffered aqueous solution
SAB1404124
Monoclonal Anti-NFIC antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution
SAB1409364
Monoclonal Anti-NFIC antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
SAB1409365
Monoclonal Anti-NFIC antibody produced in mouse, clone 2C3, ascites fluid, solution
SAB1409366
Monoclonal Anti-NFIC antibody produced in mouse, clone 2B6, ascites fluid, solution
T5648
Tamoxifen, ≥99%
C26H29NO