EMAIL THIS PAGE TO A FRIEND
Cancer research
The Neuronal Pentraxin-2 Pathway Is an Unrecognized Target in Human Neuroblastoma, Which Also Offers Prognostic Value in Patients.
PMID 26294210
Abstract
Neuronal pentraxins (NPTX) and their corresponding receptors (NPTXR) have been studied as synapse-associated proteins in the nervous system, but their role in cancer is largely unknown. By applying a multidisciplinary, high-throughput proteomic approach, we have recently identified a peptide ligand motif for targeted drug delivery to neuroblastoma. Here, we report the sequence similarity between this peptide and a conserved portion of the pentraxin domain that is involved in the homo- and hetero-oligomerization of NPTX2 and NPTXR. We show that, in comparison with normal tissues, NPTX2 and NPTXR are overexpressed in vivo in mouse models, as well as in human Schwannian stroma-poor, stage IV neuroblastoma. Both proteins are concentrated in the vicinity of tumor blood vessels, with NPTXR also present on neuroblastic tumor cells. In vivo targeting of NPTX2 and NPTXR with the selected peptide or with specific antibodies reduces tumor burden in orthotopic mouse models of human neuroblastoma. In vitro interference with this ligand/receptor system inhibits the organization of neuroblastoma cells in tumor-like masses in close contact with vascular cells, as well as their adhesion to normal microenvironment-derived cells, suggesting a role in the cross-talk between tumor and normal cells in the early steps of neuroblastoma development. Finally, we show that NPTX2 is a marker of poor prognosis for neuroblastoma patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3118
CPNQ, ≥98% (HPLC), solid
C20H17ClN4O3