EMAIL THIS PAGE TO A FRIEND
BioMed research international
Mmu-miR-27a-5p-Dependent Upregulation of MCPIP1 Inhibits the Inflammatory Response in LPS-Induced RAW264.7 Macrophage Cells.
PMID 26295043
Abstract
Lipopolysaccharide (LPS) stimulates macrophages to release proinflammatory cytokines. MicroRNAs (miRNAs) are short noncoding RNAs that are involved in inflammatory reaction. Our previously study identified the downregulated expression of mmu-miR-27a-5p in RAW267.4 cells treated with LPS. To dissect the mechanism that mmu-miR-27a-5p regulates target genes and affects proinflammatory cytokine secretion more clearly, based on previous bioinformatics prediction data, one of the potential target genes, MCPIP1 was observed to be upregulated with qRT-PCR and western blot. Luciferase reporter assays were performed to further confirm in silico prediction and determine that MCPIP1 is the target of mmu-miR-27-5p. The results suggested that mmu-miR-27a-5p directly targeted the 3'-UTR of MCPIP1 and the interaction between mmu-miR-27-5p and the 3'-UTR of MCPIP1 is sequence-specific. MCPIP1 overexpression decreased the secretion of IL-6, IL-1β, and IL-10 in macrophage cells stimulated with LPS. Our findings may provide the important information for the precise roles of mmu-miR-27a-5p in the macrophage inflammatory response to LPS stimulation in the future.