EMAIL THIS PAGE TO A FRIEND
Clinical laboratory
Performance Evaluation of the Sysmex CS-5100 Automated Coagulation Analyzer.
PMID 26299062
Abstract
Coagulation testing is widely applied clinically, and laboratories increasingly demand automated coagulation analyzers with short turn-around times and high-throughput. The purpose of this study was to evaluate the performance of the Sysmex CS-5100 automated coagulation analyzer for routine use in a clinical laboratory. The prothrombin time (PT), international normalized ratio (INR), activated partial thromboplastin time (APTT), fibrinogen (Fbg), and D-dimer were compared between the Sysmex CS-5100 and Sysmex CA-7000 analyzers, and the imprecision, comparison, throughput, STAT function, and performance for abnormal samples were measured in each. The within-run and between-run coefficients of variation (CV) for the PT, APTT, INR, and D-dimer analyses showed excellent results both in the normal and pathologic ranges. The correlation coefficients between the Sysmex CS-5100 and Sysmex CA-7000 were highly correlated. The throughput of the Sysmex CS-5100 was faster than that of the Sysmex CA-7000. There was no interference at all by total bilirubin concentrations and triglyceride concentrations in the Sysmex CS-5100 analyzer. We demonstrated that the Sysmex CS-5100 performs with satisfactory imprecision and is well suited for coagulation analysis in laboratories processing large sample numbers and icteric and lipemic samples.