EMAIL THIS PAGE TO A FRIEND
Monoclonal antibodies in immunodiagnosis and immunotherapy
Preparation and Characterization of a Monoclonal Antibody Against Morphine.
PMID 26301931
Abstract
A monoclonal antibody (MAb) was produced by immunization of a BALB/c mouse with a conjugated morphine C6-hemisuccinated derivative (MHS) to cationized bovine serum albumin (cBSA). The hybridoma clones were screened by indirect ELISA using MHS-BSA. The best hybridoma clone was subcloned thrice by limiting dilution. This hybridoma was found to be of IgG2b class and subclass and contained lambda light chain. The affinity of the MAb to morphine was obtained 2.8×10(9) M(-1). The titer of the cell culture supernatant was at least 1:800. The MAb was cross-reacted with codeine (100%) and apomorphine (16.5%), but not with heroin, naloxone, naltrexone, or papaverine. Morphine was conjugated to HRP using a mixed anhydride method and a direct competitive ELISA was designed using anti-morphine MAb. The assay was sensitive over the 50 ng/mL to 5 μg/mL concentration range. In conclusion, this MAb is useful for the development of immunoassays to measure morphine in urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284505
1,2-Dichloroethane, anhydrous, 99.8%
C2H4Cl2
07-2101
1,2-Dichloroethane, for HPLC
C2H4Cl2
C-006
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material
C18H21NO3
C-015
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
C18H21NO3
195804
Ethylenediamine dihydrochloride, 98%
C2H8N2 · 2HCl
177989
Isobutyl chloroformate, 98%
C5H9ClO2
L5626
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
C6H14N2O2 · HCl
L8662
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H14N2O2 · HCl
L7039
L-Lysine monohydrochloride, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H14N2O2 · HCl
62929
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
C6H14N2O2 · HCl
T0886
Triethylamine, ≥99%
C6H15N
90335
Triethylamine, BioUltra, ≥99.5% (GC)
C6H15N
90337
Triethylamine, for amino acid analysis, ≥99.5% (GC)
C6H15N
90338
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
C6H15N
90340
Triethylamine, puriss. p.a., ≥99.5% (GC)
C6H15N
16304
Triethylamine, purum, ≥99% (GC)
C6H15N
471283
Triethylamine, ≥99.5%
C6H15N
W424601
Triethylamine, ≥99.5%
C6H15N