PloS one
Genome-Wide Profiling of PARP1 Reveals an Interplay with Gene Regulatory Regions and DNA Methylation.
PMID 26305327
Abstract
Poly (ADP-ribose) polymerase-1 (PARP1) is a nuclear enzyme involved in DNA repair, chromatin remodeling and gene expression. PARP1 interactions with chromatin architectural multi-protein complexes (i.e. nucleosomes) alter chromatin structure resulting in changes in gene expression. Chromatin structure impacts gene regulatory processes including transcription, splicing, DNA repair, replication and recombination. It is important to delineate whether PARP1 randomly associates with nucleosomes or is present at specific nucleosome regions throughout the cell genome. We performed genome-wide association studies in breast cancer cell lines to address these questions. Our studies show that PARP1 associates with epigenetic regulatory elements genome-wide, such as active histone marks, CTCF and DNase hypersensitive sites. Additionally, the binding of PARP1 to chromatin genome-wide is mutually exclusive with DNA methylation pattern suggesting a functional interplay between PARP1 and DNA methylation. Indeed, inhibition of PARylation results in genome-wide changes in DNA methylation patterns. Our results suggest that PARP1 controls the fidelity of gene transcription and marks actively transcribed gene regions by selectively binding to transcriptionally active chromatin. These studies provide a platform for developing our understanding of PARP1's role in gene regulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P248 Monoclonal Anti-PARP antibody produced in mouse, clone C-2-10, ascites fluid
WH0000142M1
Monoclonal Anti-PARP1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
SAB1403527
Monoclonal Anti-PARP1, (N-terminal) antibody produced in mouse, clone 2C7, purified immunoglobulin, buffered aqueous solution
PLA0184
Rabbit anti-PARP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
S4264
Spermine, amorphous semi-solid, BioReagent, suitable for cell culture
C10H26N4
S3256
Spermine, ≥97%
C10H26N4
85590
Spermine, ≥99.0% (GC)
C10H26N4
302643
Trypan Blue, Dye content 60 %
C34H24N6Na4O14S4
T6146
Trypan Blue, powder, BioReagent, suitable for cell culture
C34H24N6Na4O14S4
T8154
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
C34H24N6Na4O14S4