PloS one
A Multiplex Assay for Detection of Staphylococcal and Streptococcal Exotoxins.
PMID 26305471
Abstract
Staphylococcal and streptococcal exotoxins, also known as superantigens, mediate a range of diseases including toxic shock syndrome, and they exacerbate skin, pulmonary and systemic infections caused by these organisms. When present in food sources they can cause enteric effects commonly known as food poisoning. A rapid, sensitive assay for the toxins would enable testing of clinical samples and improve surveillance of food sources. Here we developed a bead-based, two-color flow cytometry assay using single protein domains of the beta chain of T cell receptors engineered for high-affinity for staphylococcal (SEA, SEB and TSST-1) and streptococcal (SpeA and SpeC) toxins. Site-directed biotinylated forms of these high-affinity agents were used together with commercial, polyclonal, anti-toxin reagents to enable specific and sensitive detection with SD50 values of 400 pg/ml (SEA), 3 pg/ml (SEB), 25 pg/ml (TSST-1), 6 ng/ml (SpeA), and 100 pg/ml (SpeC). These sensitivities were in the range of 4- to 80-fold higher than achieved with standard ELISAs using the same reagents. A multiplex format of the assay showed reduced sensitivity due to higher noise associated with the use of multiple polyclonal agents, but the sensitivities were still well within the range necessary for detection in food sources or for rapid detection of toxins in culture supernatants. For example, the assay specifically detected toxins in supernatants derived from cultures of Staphylococcus aureus. Thus, these reagents can be used for simultaneous detection of the toxins in food sources or culture supernatants of potential pathogenic strains of Staphylococcus aureus and Streptococcus pyogenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700855 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700857 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700851 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700853 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700847 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700859 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Texas Red® antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700854 Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700856 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700850 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700852 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700846 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700858 Anti-Rabbit IgG (Fc specific)-Texas Red® antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600006 Anti-Rabbit IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600015
Anti-Rabbit IgG (H+L), CF 350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600021
Anti-Rabbit IgG (H+L), CF 405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600024
Anti-Rabbit IgG (H+L), CF 405S antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600044
Anti-Rabbit IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600068
Anti-Rabbit IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600084
Anti-Rabbit IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600107
Anti-Rabbit IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600140
Anti-Rabbit IgG (H+L), CF 633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600163
Anti-Rabbit IgG (H+L), CF 640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600184
Anti-Rabbit IgG (H+L), CF 647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600193
Anti-Rabbit IgG (H+L), CF 660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600195
Anti-Rabbit IgG (H+L), CF 660R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600212
Anti-Rabbit IgG (H+L), CF 750 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600223
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600406
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600389
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600300
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution