EMAIL THIS PAGE TO A FRIEND
Genes, chromosomes & cancer
Methyltransferase expression and tumor suppressor gene methylation in sporadic and familial colorectal cancer.
PMID 26305882
Abstract
Molecular mechanisms underlying coordinated hypermethylation of multiple CpG islands in cancer remain unclear and studies of methyltransferase enzymes have arrived at conflicting results. We focused on DNMT1 and DNMT3B, DNA methyltransferases responsible for (de novo) methylation, and EZH2, histone (H3K27) methyltransferase, and examined their roles in tumor suppressor gene (TSG) methylation patterns we have previously established in sporadic and familial cancers. Our investigation comprised 165 tumors, stratified by tissue of origin (117 colorectal and 48 endometrial carcinomas) and sporadic vs. familial disease (57 sporadic vs. 60 familial, mainly Lynch syndrome, colorectal carcinomas). By immunohistochemical evaluation, EZH2 protein expression was associated with a TSG methylator phenotype. DNMT1, DNMT3B, and EZH2 were expressed at significantly higher levels in tumor vs. normal tissues. DNMT1 and EZH2 expression were positively correlated and higher in microsatellite-unstable vs. microsatellite-stable tumors, whether sporadic or hereditary. Ki-67 expression mirrored the same pattern. Promoter methylation of the methyltransferase genes themselves was addressed as a possible cause behind their altered expression. While DNMT1 or EZH2 did not show differential methylation between normal and tumor tissues, DNMT3B analysis corroborated the regulatory role of a distal promoter region. Our study shows that methyltransferase expression in cancer depends on the tissue of origin, microsatellite-instability status, cellular proliferation, and--in the case of DNMT3B--promoter methylation of the respective gene. Translation of methyltransferase expression into DNA methylation appears complex as suggested by the fact that except for EZH2, no clear association between methyltransferase protein expression and TSG methylation was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS3805
Anti-AIM (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3807
Anti-AIM (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22659 Anti-DNMT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500321
Anti-DNMT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
D4692
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D4567
Anti-DNMT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA002694 Anti-DNMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106406
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody
SAB2700189
Anti-Dnmt3b antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001595
Anti-DNMT3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E7031
Anti-EZH2 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405776
Anti-EZH2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
E6906
Anti-EZH2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV38470
Anti-EZH2 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2106436
Anti-EZH2 antibody produced in rabbit, affinity isolated antibody
SAB1410354
Anti-EZH2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1403742
Monoclonal Anti-DNMT1, (N-terminal) antibody produced in mouse, clone 2B5, ascites fluid
SAB1408985 Monoclonal Anti-EZH2 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
PLA0011
Rabbit anti-DNMT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
T1952 Trichostatin A, Ready Made Solution, 5 mM in DMSO (0.2 μm-filtered), from Streptomyces sp.
C17H22N2O3