EMAIL THIS PAGE TO A FRIEND
PLoS genetics
SmD1 Modulates the miRNA Pathway Independently of Its Pre-mRNA Splicing Function.
PMID 26308709
Abstract
microRNAs (miRNAs) are a class of endogenous regulatory RNAs that play a key role in myriad biological processes. Upon transcription, primary miRNA transcripts are sequentially processed by Drosha and Dicer ribonucleases into ~22-24 nt miRNAs. Subsequently, miRNAs are incorporated into the RNA-induced silencing complexes (RISCs) that contain Argonaute (AGO) family proteins and guide RISC to target RNAs via complementary base pairing, leading to post-transcriptional gene silencing by a combination of translation inhibition and mRNA destabilization. Select pre-mRNA splicing factors have been implicated in small RNA-mediated gene silencing pathways in fission yeast, worms, flies and mammals, but the underlying molecular mechanisms are not well understood. Here, we show that SmD1, a core component of the Drosophila small nuclear ribonucleoprotein particle (snRNP) implicated in splicing, is required for miRNA biogenesis and function. SmD1 interacts with both the microprocessor component Pasha and pri-miRNAs, and is indispensable for optimal miRNA biogenesis. Depletion of SmD1 impairs the assembly and function of the miRISC without significantly affecting the expression of major canonical miRNA pathway components. Moreover, SmD1 physically and functionally associates with components of the miRISC, including AGO1 and GW182. Notably, miRNA defects resulting from SmD1 silencing can be uncoupled from defects in pre-mRNA splicing, and the miRNA and splicing machineries are physically and functionally distinct entities. Finally, photoactivatable-ribonucleoside-enhanced crosslinking and immunoprecipitation (PAR-CLIP) analysis identifies numerous SmD1-binding events across the transcriptome and reveals direct SmD1-miRNA interactions. Our study suggests that SmD1 plays a direct role in miRNA-mediated gene silencing independently of its pre-mRNA splicing activity and indicates that the dual roles of splicing factors in post-transcriptional gene regulation may be evolutionarily widespread.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA021289
Anti-BBS9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408674
Anti-BBS9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1304992 ANTI-DIO1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2101188
Anti-JAG2 antibody produced in rabbit, affinity isolated antibody
HPA030636 Anti-JAG2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407254
Anti-LMOD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101354
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB4200200
Anti-LMOD1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA028325
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028435
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030097
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB2107079
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB1305785
ANTI-PTHB1(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1402173 Monoclonal Anti-DIO1 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
WH0003714M3
Monoclonal Anti-JAG2 antibody produced in mouse, clone 4F10, purified immunoglobulin, buffered aqueous solution
SAB1403998
Monoclonal Anti-JAG2 antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
SAB1409259 Monoclonal Anti-JAG2 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB4200496
Monoclonal Anti-LMOD1 antibody produced in mouse, ~1.0 mg/mL, clone LMOD1-12, purified immunoglobulin
SAB1401605
Monoclonal Anti-LMOD1 antibody produced in mouse, clone 3B8, purified immunoglobulin, buffered aqueous solution