EMAIL THIS PAGE TO A FRIEND
Acta biomaterialia
Intrinsic and extrinsic determinants of central nervous system axon outgrowth into alginate-based anisotropic hydrogels.
PMID 26310676
Abstract
Appropriate target reinnervation and functional recovery after spinal cord injury depend on longitudinally directed regrowth of injured axons. Anisotropic alginate-based capillary hydrogels (ACH) support peripheral nervous system derived axon growth, which is accompanied by glial supporting cell migration into the ACH. The aim of the present study was to analyze central nervous system (CNS) derived (entorhinal cortex, spinal cord slice cultures) axon regrowth into ACH containing linearly aligned capillaries of defined capillary sizes without and with gelatin constituent. Anisotropic ACH were prepared by ionotropic gel formation using Ba(2+), Cu(2+), Sr(2+), or Zn(2+) ions resulting in gels with average capillary diameters of 11, 13, 29, and 89μm, respectively. Postnatal rat entorhinal cortex or spinal cord slice cultures were placed on top of 500μm thick ACH. Seven days later axon growth and astroglial migration into the ACH were determined. Axon density within capillaries correlated positively with increasing capillary diameters, whereas longitudinally oriented axon outgrowth diminished with increasing capillary diameter. Axons growing into the hydrogels were always accompanied by astrocytes strongly suggesting that respective cells are required to mediate CNS axon elongation into ACH. Overall, midsize capillary diameter ACH appeared to be the best compromise between axon density and orientation. Taken together, ACH promote CNS axon ingrowth, which is determined by the capillary diameter and migration of slice culture derived astroglia into the hydrogel. Biomaterials are investigated as therapeutic tools to bridge irreversible lesions following traumatic spinal cord injury. The goal is to develop biomaterials, which promote longitudinally oriented regeneration of as many injured axons as possible as prerequisite for substantial functional recovery. Optimal parameters of the biomaterial have yet to be defined. In the present study we show that increasing capillary diameters within such hydrogels enhanced central nervous system axon regeneration at the expense of longitudinal orientation. Axon ingrowth into the hydrogels was only observed in the presence of glial supporting cells, namely astrocytes. This suggests that alginate-based hydrogels need to be colonized with respective cells in order to facilitate axon ingrowth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76864
4-(4,5-Diphenyl-2-imidazolyl)phenol, suitable for chemiluminescence, ≥99.0% (HPLC)
C21H16N2O
00561
Acetone, for luminescence, ≥99.5% (GC)
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O
W332615
Acetone, natural, ≥97%
C3H6O
01-0520
Acetone, for HPLC
C3H6O
01-0494
Acetone, for residue analysis, suitable for 5000 per JIS
C3H6O
01-0475
Acetone, suitable for determination of dioxins
C3H6O
48358
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C3H6O
686530
Acetone solution, contains 20.0 % (v/v) acetonitrile, for HPLC
C3H6O
52649
Hexamethylene diisocyanate, puriss., ≥99.0% (GC)
C8H12N2O2
225711
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
HNO3
40230
Nitric acid, semiconductor grade PURANAL (Honeywell 17818), ≥65%
HNO3
40222
Nitric acid, semiconductor grade PURANAL (Honeywell 17952), ≥69%
HNO3
40280
Nitric acid, semiconductor grade MOS PURANAL (Honeywell 17920), ≥69%
HNO3
695025
Nitric acid, reagent grade, fuming, >90%
HNO3
695041
Nitric acid, ACS reagent, ≥90.0%
HNO3
608769
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
H14NO3
311421
Perchloric acid, 70%, 99.999% trace metals basis
HClO4
244252
Perchloric acid, ACS reagent, 70%
HClO4
311413
Perchloric acid, ACS reagent, 60%
HClO4
77234
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
HClO4
77228
Perchloric acid, puriss. p.a., ACS reagent, 70% (Hg ≤0.0000005%), packed in coated, shock- and leak-protected glass bottle, ≥69% (T)
HClO4
30755
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
HClO4
P6061
Phenylacetic acid, plant cell culture tested
C8H8O2
P16621
Phenylacetic acid, 99%
C8H8O2
W287814
Phenylacetic acid, natural, ≥99%
C8H8O2
W287806
Phenylacetic acid, ≥99%, FCC, FG
C8H8O2
R2751
Ruthenium Red, Technical grade
H42Cl6N14O2Ru3
84071
Ruthenium Red, for microscopy, ≥85% (calc. on dry substance, AT)
H42Cl6N14O2Ru3