EMAIL THIS PAGE TO A FRIEND
Clinical and vaccine immunology : CVI
Generation and Efficacy Evaluation of a Recombinant Pseudorabies Virus Variant Expressing the E2 Protein of Classical Swine Fever Virus in Pigs.
PMID 26311244
Abstract
Classical swine fever (CSF) is an economically important infectious disease of pigs caused by classical swine fever virus (CSFV). Pseudorabies (PR), which is caused by pseudorabies virus (PRV), is another important infectious disease of pigs and other animals. Coinfections of pigs with PRV and CSFV occur occasionally in the field. The modified live vaccine Bartha-K61 strain has played an important role in the control of PR in many countries, including China. Since late 2011, however, increasing PR outbreaks caused by an emerging PRV variant have been reported in Bartha-K61-vaccinated swine populations on many farms in China. Previously, we generated a gE/gI-deleted PRV (rPRVTJ-delgE) based on this PRV variant, which was shown to be safe and can provide rapid and complete protection against lethal challenge with the PRV variant in pigs. Here, we generated a new recombinant PRV variant expressing the E2 gene of CSFV (rPRVTJ-delgE/gI-E2) and evaluated its immunogenicity and efficacy in pigs. The results showed that rPRVTJ-delgE/gI-E2 was safe for pigs, induced detectable anti-PRV and anti-CSFV neutralizing antibodies, and provided complete protection against the lethal challenge with either the PRV TJ strain or the CSFV Shimen strain. The data indicate that rPRVTJ-delgE/gI-E2 is a promising candidate bivalent vaccine against PRV and CSFV coinfections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22284F
Anti-ATP6V1E1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1401024
Anti-ATP6V1E1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA029196
Anti-ATP6V1E1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400071
Anti-DBT antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA026481
Anti-DBT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA026485
Anti-DBT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA026533
Anti-DBT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
SAB2104091
Anti-DBT, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22844 Anti-DLAT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1408946
Anti-DLAT antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV41994
Anti-DLAT antibody produced in rabbit, IgG fraction of antiserum
HPA040786
Anti-DLAT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410301
Anti-DLAT antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400079
Anti-DLST antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2701694
Anti-dlst antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003010
Anti-DLST antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401116
Anti-DLST antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2701135
Anti-DLST antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1402699
Monoclonal Anti-ATP6V1E1 antibody produced in mouse, clone 4E11, purified immunoglobulin, buffered aqueous solution
SAB1412704
Monoclonal Anti-DLST antibody produced in mouse, clone 4D7, purified immunoglobulin, 1 mg/mL