EMAIL THIS PAGE TO A FRIEND
Nature communications
PAQR3 modulates cholesterol homeostasis by anchoring Scap/SREBP complex to the Golgi apparatus.
PMID 26311497
Abstract
Cholesterol biosynthesis is regulated by transcription factors SREBPs and their escort protein Scap. On sterol depletion, Scap/SREBP complex is transported from endoplasmic reticulum (ER) to the Golgi apparatus where SREBP is activated. Under cholesterol sufficient condition, Insigs act as anchor proteins to retain Scap/SREBP in the ER. However, the anchor protein of Scap/SREBP in the Golgi is unknown. Here we report that a Golgi-localized membrane protein progestin and adipoQ receptors 3 (PAQR3) interacts with Scap and SREBP and tethers them to the Golgi. PAQR3 promotes Scap/SREBP complex formation, potentiates SREBP processing and enhances lipid synthesis. The mutually exclusive interaction between Scap and PAQR3 or Insig-1 is regulated by cholesterol level. PAQR3 knockdown in liver blunts SREBP pathway and decreases hepatic cholesterol content. Disrupting the interaction of PAQR3 with Scap/SREBP by a synthetic peptide inhibits SREBP processing and activation. Thus, PAQR3 regulates cholesterol homeostasis by anchoring Scap/SREBP to the Golgi and disruption of such function reduces cholesterol biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21184P Anti-c-Myc (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21222 Anti-c-Myc (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C4731
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
C7617
Anti-Calnexin antibody, Mouse monoclonal, clone TO-5, purified from hybridoma cell culture
SAB4503258
Anti-Calnexin, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501291
Anti-CANX antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA009433
Anti-CANX antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA009696
Anti-CANX antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5200084
Anti-CANX antibody produced in rabbit, whole antiserum
SAB5200097 Anti-CANX antibody produced in rabbit, affinity isolated antibody
G7295
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1409131
Anti-GOLGA1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2107066
Anti-GOLGA1 antibody produced in rabbit, affinity isolated antibody
SAB1410558
Anti-GOLGA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1405886
Anti-GOLGA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021178
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021230
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA021799
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
SAB2101165
Anti-INSIG1 (ab1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2101166
Anti-INSIG1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2103102
Anti-INSIG1 antibody produced in rabbit, affinity isolated antibody
SAB2104446
Anti-INSIG1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA005494
Anti-MBTPS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502122
Anti-MBTPS2 antibody produced in rabbit, affinity isolated antibody
SAB3701101 Anti-Mouse IgG (H+L) antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB4300319
Anti-MYC (Ab-358) antibody produced in rabbit, affinity isolated antibody