EMAIL THIS PAGE TO A FRIEND
Journal of virology
Knockout of cGAS and STING Rescues Virus Infection of Plasmid DNA-Transfected Cells.
PMID 26311870
Abstract
It is well known that plasmid DNA transfection, prior to virus infection, negatively affects infection efficiency. Here, we show that cytosolic plasmid DNA activates the cGAS/STING signaling pathway, which ultimately leads to the induction of an antiviral state of the cells. Using a transient one-plasmid clustered regularly interspaced short palindromic repeat (CRISPR)/Cas9 system, we generated cGAS/STING-knockout cells and show that these cells can be infected after plasmid DNA transfection as efficiently as nontransfected cells.